MilliporeSigma
Signature microRNAs in human cornea limbal epithelium.
Functional & integrative genomics (2014-12-10)
Yufei Teng, Hoi Kin Wong, Vishal Jhanji, Jian Huan Chen, Alvin Lerrmann Young, Mingzhi Zhang, Kwong Wai Choy, Jodhbir Singh Mehta, Chi Pui Pang, Gary Hin-Fai Yam
ABSTRACT
This study was aimed to identify the signature microRNAs, which regulate the biological processes of corneal epithelial progenitor cell (CEPC) homeostasis and regulation through characterizing the differential expression profile of microRNAs in human limbal epithelium containing adult CEPC versus central corneal epithelium without CEPC. MicroRNA microarray had identified 37 microRNAs enriched in human corneal epithelium. Among them, nine were significantly upregulated in limbal epithelium and one in central corneal epithelium after validation by TaqMan® real-time polymerase chain reaction. In addition to our previous finding of miR-143 and 145, the expression of miR-10b, 126, and 155 was localized in limbal epithelium (LE) (predominantly basal layers) by using locked nucleic acid-based in situ hybridization. Potential target genes were predicted by TargetScan Human v6.0 and compared to the reported human cornea epithelial gene profile GSE5543. Analyzed by web-based Kyoto Encyclopedia of Genes and Genomes (KEGG) pathway and DAVID Functional Annotation Bioinformatics Resources v6.7, the downregulated genes were involved in pathways of immune response and cellular protection, apoptosis, and cell movement whereas upregulated genes with cell survival, cell-matrix interaction, and cell-cell adhesion. We found a constant occurrence of miR-143, 145, and 155 in all KEGG pathways regulating limbal epithelial events. By Ingenuity Systems (IPA®) analysis, these microRNAs could cooperatively regulate cell growth and apoptosis via tumor necrosis factor activation and MYC repression. Our findings thus suggest a unique microRNA signature existing in human limbal epithelium and participating in CEPC homeostasis.
Sigma-Aldrich
Chloroform, suitable for HPLC
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Estimated to ship on March 23, 2025
MXP 2,865.00
100 g
Estimated to ship on March 23, 2025
MXP 8,909.00
500 g
Estimated to ship on March 23, 2025
MXP 19,563.00
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Estimated to ship on March 23, 2025
MXP 2,865.00
100 g
Estimated to ship on March 23, 2025
MXP 8,909.00
500 g
Estimated to ship on March 23, 2025
MXP 19,563.00
Sigma-Aldrich
Chloroform, SAJ super special grade, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Estimated to ship on March 23, 2025
MXP 2,865.00
100 g
Estimated to ship on March 23, 2025
MXP 8,909.00
500 g
Estimated to ship on March 23, 2025
MXP 19,563.00
Sigma-Aldrich
Chloroform, SAJ first grade, ≥99.0%, contains 0.4-0.8% ethanol
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Estimated to ship on March 23, 2025
MXP 2,865.00
100 g
Estimated to ship on March 23, 2025
MXP 8,909.00
500 g
Estimated to ship on March 23, 2025
MXP 19,563.00
Sigma-Aldrich
Chloroform, JIS 300, ≥99.0%, for residue analysis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Estimated to ship on March 23, 2025
MXP 2,865.00
100 g
Estimated to ship on March 23, 2025
MXP 8,909.00
500 g
Estimated to ship on March 23, 2025
MXP 19,563.00
Sigma-Aldrich
Chloroform, JIS special grade, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Estimated to ship on March 23, 2025
MXP 2,865.00
100 g
Estimated to ship on March 23, 2025
MXP 8,909.00
500 g
Estimated to ship on March 23, 2025
MXP 19,563.00
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Estimated to ship on March 23, 2025
MXP 2,865.00
100 g
Estimated to ship on March 23, 2025
MXP 8,909.00
500 g
Estimated to ship on March 23, 2025
MXP 19,563.00
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Estimated to ship on March 23, 2025
MXP 2,865.00
100 g
Estimated to ship on March 23, 2025
MXP 8,909.00
500 g
Estimated to ship on March 23, 2025
MXP 19,563.00
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Estimated to ship on March 23, 2025
MXP 2,865.00
100 g
Estimated to ship on March 23, 2025
MXP 8,909.00
500 g
Estimated to ship on March 23, 2025
MXP 19,563.00
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Estimated to ship on March 23, 2025
MXP 2,865.00
100 g
Estimated to ship on March 23, 2025
MXP 8,909.00
500 g
Estimated to ship on March 23, 2025
MXP 19,563.00
Supelco
Chloroform, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Estimated to ship on March 23, 2025
MXP 2,865.00
100 g
Estimated to ship on March 23, 2025
MXP 8,909.00
500 g
Estimated to ship on March 23, 2025
MXP 19,563.00
Trolamine, European Pharmacopoeia (EP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Estimated to ship on March 23, 2025
MXP 2,865.00
100 g
Estimated to ship on March 23, 2025
MXP 8,909.00
500 g
Estimated to ship on March 23, 2025
MXP 19,563.00
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Estimated to ship on March 23, 2025
MXP 2,865.00
100 g
Estimated to ship on March 23, 2025
MXP 8,909.00
500 g
Estimated to ship on March 23, 2025
MXP 19,563.00
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Estimated to ship on March 23, 2025
MXP 2,865.00
100 g
Estimated to ship on March 23, 2025
MXP 8,909.00
500 g
Estimated to ship on March 23, 2025
MXP 19,563.00
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Estimated to ship on March 23, 2025
MXP 2,865.00
100 g
Estimated to ship on March 23, 2025
MXP 8,909.00
500 g
Estimated to ship on March 23, 2025
MXP 19,563.00
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, amylene stabilized
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Estimated to ship on March 23, 2025
MXP 2,865.00
100 g
Estimated to ship on March 23, 2025
MXP 8,909.00
500 g
Estimated to ship on March 23, 2025
MXP 19,563.00
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Estimated to ship on March 23, 2025
MXP 2,865.00
100 g
Estimated to ship on March 23, 2025
MXP 8,909.00
500 g
Estimated to ship on March 23, 2025
MXP 19,563.00
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Estimated to ship on March 23, 2025
MXP 2,865.00
100 g
Estimated to ship on March 23, 2025
MXP 8,909.00
500 g
Estimated to ship on March 23, 2025
MXP 19,563.00
Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Estimated to ship on March 23, 2025
MXP 2,865.00
100 g
Estimated to ship on March 23, 2025
MXP 8,909.00
500 g
Estimated to ship on March 23, 2025
MXP 19,563.00
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Estimated to ship on March 23, 2025
MXP 2,865.00
100 g
Estimated to ship on March 23, 2025
MXP 8,909.00
500 g
Estimated to ship on March 23, 2025
MXP 19,563.00
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Estimated to ship on March 23, 2025
MXP 2,865.00
100 g
Estimated to ship on March 23, 2025
MXP 8,909.00
500 g
Estimated to ship on March 23, 2025
MXP 19,563.00
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Estimated to ship on March 23, 2025
MXP 2,865.00
100 g
Estimated to ship on March 23, 2025
MXP 8,909.00
500 g
Estimated to ship on March 23, 2025
MXP 19,563.00
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Estimated to ship on March 23, 2025
MXP 2,865.00
100 g
Estimated to ship on March 23, 2025
MXP 8,909.00
500 g
Estimated to ship on March 23, 2025
MXP 19,563.00
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Estimated to ship on March 23, 2025
MXP 2,865.00
100 g
Estimated to ship on March 23, 2025
MXP 8,909.00
500 g
Estimated to ship on March 23, 2025
MXP 19,563.00
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Estimated to ship on March 23, 2025
MXP 2,865.00
100 g
Estimated to ship on March 23, 2025
MXP 8,909.00
500 g
Estimated to ship on March 23, 2025
MXP 19,563.00
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Estimated to ship on March 23, 2025
MXP 2,865.00
100 g
Estimated to ship on March 23, 2025
MXP 8,909.00
500 g
Estimated to ship on March 23, 2025
MXP 19,563.00
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Estimated to ship on March 23, 2025
MXP 2,865.00
100 g
Estimated to ship on March 23, 2025
MXP 8,909.00
500 g
Estimated to ship on March 23, 2025
MXP 19,563.00
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Estimated to ship on March 23, 2025
MXP 2,865.00
100 g
Estimated to ship on March 23, 2025
MXP 8,909.00
500 g
Estimated to ship on March 23, 2025
MXP 19,563.00
Sigma-Aldrich
Triethanolamine, BioUltra, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Estimated to ship on March 23, 2025
MXP 2,865.00
100 g
Estimated to ship on March 23, 2025
MXP 8,909.00
500 g
Estimated to ship on March 23, 2025
MXP 19,563.00
Sigma-Aldrich
Triethanolamine, puriss. p.a., ≥99% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Estimated to ship on March 23, 2025
MXP 2,865.00
100 g
Estimated to ship on March 23, 2025
MXP 8,909.00
500 g
Estimated to ship on March 23, 2025
MXP 19,563.00