MilliporeSigma
  • Hydrogels as feeder-free scaffolds for long-term self-renewal of mouse induced pluripotent stem cells.
Hydrogels as feeder-free scaffolds for long-term self-renewal of mouse induced pluripotent stem cells.
Journal of tissue engineering and regenerative medicine (2012-11-21)
Jing Jing Yang, Jian Fang Liu, Takayuki Kurokawa, Kazuhiro Kitada, Jian Ping Gong
ABSTRACT
Expanding undifferentiated induced pluripotent stem (iPS) cells in vitro is a basic requirement for application of iPS cells in both fundamental research and clinical regeneration. In this study, we intended to establish a simple, low cost and efficient method for the long-term self-renewal of mouse induced pluripotent stem (miPS) cells without using feeder-cells and adhesive proteins. Three scaffolds were selected for the long-term subculture of miPS cells over two months starting from passages 14 to 29: 1) a gelatin coated polystyrene (Gelatin-PS) that is a widely used scaffold for self-renewal of mouse embryonic stem (mES) cells; 2) a neutral hydrogel poly(N,N-dimethylacrylamide) (PDMAAm); and 3) a negatively charged hydrogel poly(2-acrylamido-2-methyl-propane sulfonic acid sodium salt) (PNaAMPS). Each passaged miPS cells on these scaffolds were cryopreserved successfully and the revived cells showed high viability and proliferation. The passaged miPS cells maintained a high undifferentiated state on all three scaffolds and a high level of pluripotency by expressing differentiation markers in vitro and forming teratomas in SCID mice with derivatives of all three germ layers. Compared to Gelatin-PS, the two hydrogels exhibited much better self-renewal performance in terms of high proliferation rate and level of expression of undifferentiated gene markers as well as efficiency in pluripotent teratoma formation. Furthermore, the PNaAMPS hydrogel demonstrated a slightly higher efficiency and simpler operation of cell expansion than the PDMAAm hydrogel. To conclude, PNaAMPS hydrogel is an excellent feeder-free scaffold because of its simplicity, low cost and high efficiency in expanding a large number of miPS cells in vitro.
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
α-Ketoglutaric acid, 99.0-101.0% (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
N,N-Dimethylacrylamide, 99%, contains 500 ppm monomethyl ether hydroquinone as inhibitor
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride solution, 0.85%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, tablet
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
α-Ketoglutaric acid, BioReagent, suitable for cell culture, suitable for insect cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride solution, 5 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
α-Ketoglutaric acid, ≥98.5% (NaOH, titration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride solution, 1 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride solution, 0.1 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SKU
Pack Size
Availability
Price
Quantity
SAFC
HEPES
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%