MilliporeSigma
  • A single-step enzyme immunoassay capillary sensor composed of functional multilayer coatings for the diagnosis of marker proteins.
A single-step enzyme immunoassay capillary sensor composed of functional multilayer coatings for the diagnosis of marker proteins.
The Analyst (2015-01-20)
Shun-ichi Funano, Masato Sugahara, Terence G Henares, Kenji Sueyoshi, Tatsuro Endo, Hideaki Hisamoto
ABSTRACT
A single-step, easy-to-use enzyme immunoassay capillary sensor, composed of functional multilayer coatings, was developed in this study. The coatings were composed of substrate-immobilized hydrophobic coating, hydrogel coating, and soluble coating containing an enzyme-labeled antibody. The response mechanism involved a spontaneous immunoreaction triggered by capillary action-mediated introduction of a sample antigen solution and subsequent separation of unreacted enzyme-labeled antibodies and antigen-enzyme-labeled antibody complexes by the molecular sieving effect of the hydrogel. An enzyme reaction at the substrate-immobilized hydrophobic coating/hydrogel coating interface resulted in a protein-selective fluorescence response. An antigen concentration-dependent response was obtained for diagnostic marker protein samples (hemoglobin A1c (HbA1c), 7.14-16.7 mg mL(-1); alpha-fetoprotein (AFP), 1.4-140 ng mL(-1); C-reactive protein (CRP), 0.5-10 μg mL(-1)) that cover a clinically important concentration range. The successful measurement of CRP in diluted serum samples demonstrated the application of this capillary sensor.
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, ≥99.0%
Sigma-Aldrich
Ammonium persulfate, JIS special grade, ≥98.0%
Sigma-Aldrich
Ammonium persulfate, SAJ first grade, ≥95.0%
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, ReagentPlus®, 99%
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, ≥99.5%, purified by redistillation
Sigma-Aldrich
Ammonium persulfate, BioUltra, for molecular biology, ≥98.0% (RT)
Supelco
Acrylamide solution, 40% in H2O, for molecular biology
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, suitable for electrophoresis, ≥99.0%
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, for molecular biology, ≥99% (GC)
Sigma-Aldrich
Ammonium persulfate, BioXtra, ≥98.0%
Sigma-Aldrich
Ammonium persulfate, for molecular biology, suitable for electrophoresis, ≥98%
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
Ammonium persulfate, ACS reagent, ≥98.0%
Sigma-Aldrich
Ammonium persulfate, reagent grade, 98%
Sigma-Aldrich
Ammonium persulfate, free-flowing, Redi-Dri, reagent grade, 98%
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, contains no stabilizer
Sigma-Aldrich
Tetrahydrofuran, JIS special grade, ≥99.5%
Sigma-Aldrich
Acrylamide, ≥98.0%
Sigma-Aldrich
Acrylamide, ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acrylamide, SAJ first grade, ≥98.0%
Supelco
Acrylamide, analytical standard
Supelco
Acrylamide, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Supelco
Tetrahydrofuran, analytical standard
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Sigma-Aldrich
Acrylamide, purum, ≥98.0% (GC)
Sigma-Aldrich
Acrylamide, for Northern and Southern blotting, powder blend
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%