MilliporeSigma
  • Triosephosphate isomerase of Taenia solium (TTPI): phage display and antibodies as tools for finding target regions to inhibit catalytic activity.
Triosephosphate isomerase of Taenia solium (TTPI): phage display and antibodies as tools for finding target regions to inhibit catalytic activity.
Parasitology research (2014-10-03)
Víctor Sanabria-Ayala, Iaraset Belmont, Landa Abraham
ABSTRACT
Previous studies demonstrated that antibodies against triosephosphate isomerase of Taenia solium (TTPI) can alter its enzymatic catalysis. In the present study, we used antibodies produced against the NH2-terminal region of TTPI (1/3NH2TTPI) and the phage display technology to find target regions to inhibit TTPI activity. As a first step, we obtained polyclonal antibodies against non-conserved regions from the 1/3NH2TTPI, which had an inhibitory effect of about 74 % on catalytic activity. Afterward, they were used to screen a library of phage-displayed dodecapeptides; as a result, 41 phage mimotope clones were isolated and grouped according to their amino acid sequence, finding the consensus A1 (VPTXPI), A2 (VPTXXI), B (LTPGQ), and D (DPLPR). Antibodies against selected phage mimotope clones were obtained by rabbit's immunization; these ones clearly recognized TTPI by both Western blot and ELISA. However, only the mimotope PDTS16 (DSVTPTSVMAVA) clone, which belongs to the VPTXXI consensus, raised antibodies capable of inhibiting the TTPI catalytic activity in 45 %. Anti-PDTS16 antibodies were confronted to several synthetic peptides that encompass the 1/3NH2TTPI, and they only recognized three, which share the motif FDTLQK belonging to the helix-α1 in TTPI. This suggests that this motif is the main part of the epitope recognized by anti-PDTS16 antibodies and revealed its importance for TTPI catalysis.
Product Description
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sulfuric acid, 99.999%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sulfuric acid, SAJ first grade, ≥95.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sulfuric acid solution, 0.05 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sulfuric acid solution, 5 mM
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sulfuric acid solution, 0.025 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sulfuric acid solution, 0.25 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sulfuric acid solution, 0.5 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sulfuric acid solution, 1.5 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sulfuric acid solution, 0.1 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sulfuric acid solution, 0.01 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sulfuric acid, JIS special grade, ≥95.0%
SKU
Pack Size
Availability
Price
Quantity
Trolamine, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Supelco
DL-Dithiothreitol solution, 1 M in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
DL-Glyceraldehyde, ≥90% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Triethanolamine, BioUltra, ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Triethanolamine, puriss. p.a., ≥99% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Triethanolamine, ≥99.0% (GC)
SKU
Pack Size
Availability
Price
Quantity
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Triethanolamine, JIS special grade, ≥98.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Triethanolamine, reagent grade, 98%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Triethanolamine, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis