MilliporeSigma
  • Synthesis, structure, spectral characterization, electrochemistry and evaluation of antibacterial potentiality of a novel oxime-based palladium(II) compound.
Synthesis, structure, spectral characterization, electrochemistry and evaluation of antibacterial potentiality of a novel oxime-based palladium(II) compound.
European journal of medicinal chemistry (2014-12-03)
Nirmalya Bandyopadhyay, Miaoli Zhu, Liping Lu, Debmalya Mitra, Mousumi Das, Piu Das, Amalesh Samanta, Jnan Prakash Naskar
ABSTRACT
The title monomeric Pd(II) compound, [Pd(L)(Cl)], was synthesized in moderate yield out of the reaction of equimolar proportion of Na2[PdCl4] and 3-[(5-bromo-2-hydroxy-benzylidene)-hydrazono]-butan-2-one oxime (LH) in tetrahydrofuran milieu. LH is a 1:1 Schiff-base condensate of 2,3-butanedionemonoxime monohydrazone and 5-bromosalicylaldehyde. [Pd(L)(Cl)] has been characterized by C, H and N microanalyses, (1)H and (13)C NMR, FAB-MS, FT-IR, Raman spectra, UV-Vis spectra and molar electrical conductivity measurements. [Pd(L)(Cl)] is diamagnetic. Structural elucidation reveals that the palladium center in [Pd(L)(Cl)] is nested in 'N2OCl' coordination environment. The geometry around Pd in [Pd(L)(Cl)] is distorted square-planar. The redox behavior of [Pd(L)(Cl)] in DMF shows a reduction couple, Pd(II)/Pd(I) at -0.836 V versus Ag/AgCl. The in vitro antimicrobial activity of [Pd(L)(Cl)] was screened against both Gram-positive and Gram-negative human pathogenic bacteria. This bioactivity was substantiated with SEM study. [Pd(L)(Cl)] exhibits satisfactory bactericidal as well as bacteriostatic activity.
Product Description
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Hydrazine hydrate solution, puriss. p.a., 24-26% in H2O (RT)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Hydrazine monohydrate, SAJ first grade, ≥98.0%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Amoxicillin, 95.0-102.0% anhydrous basis
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Supelco
3-Nitrobenzyl alcohol, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Hydrazine monohydrate, N2H4 64-65 %, reagent grade, ≥97%
Sigma-Aldrich
3-Nitrobenzyl alcohol, 98%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology