MilliporeSigma
  • Characterization of pressurized hot water extracts of grape pomace: chemical and biological antioxidant activity.
Characterization of pressurized hot water extracts of grape pomace: chemical and biological antioxidant activity.
Food chemistry (2014-10-14)
J R Vergara-Salinas, Mauricio Vergara, Claudia Altamirano, Álvaro Gonzalez, J R Pérez-Correa
ABSTRACT
Pressurized hot water extracts obtained at different temperatures possess different compositions and antioxidant activities and, consequently, different bioactivities. We characterized two pressurized hot water extracts from grape pomace obtained at 100°C (GPE100) and 200°C (GPE200) in terms of antioxidant activity and composition, as well as protective effect on cell growth and mitochondrial membrane potential (Δψm) in a HL-60 cell culture under oxidative conditions. GPE100 extracts were richer in polyphenols and poorer in Maillard reaction products (MRPs) than were GPE200 extracts. Moreover, hydroxymethylfurfural was detected only in GPE200. Both extracts exhibited similar protective effects on cell growth (comparable to the effect of trolox). In addition, GPE100 strongly decreased the Δψm loss, reaching values even lower than those of the control culture. This protective effect may be related to its high polyphenols content. At the highest concentration assessed, both extracts showed strong cytotoxicity, especially GPE200. This cytotoxicity could be related to their MRPs content.
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Pricing and availability is not currently available.
Supelco
2,4,6-Tris(2-pyridyl)-s-triazine, for spectrophotometric det. of Fe, ≥99.0% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Pricing and availability is not currently available.
Sigma-Aldrich
5-(Hydroxymethyl)furfural, ≥99%, FG
Pricing and availability is not currently available.
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Pricing and availability is not currently available.
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Pricing and availability is not currently available.
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
5-Hydroxymethyl-2-furaldehyde, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Pricing and availability is not currently available.
Supelco
L-Ascorbic acid, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Pricing and availability is not currently available.
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Pricing and availability is not currently available.