MilliporeSigma
  • Development and validation of a GC/MS method for the simultaneous determination of levetiracetam and lamotrigine in whole blood.
Development and validation of a GC/MS method for the simultaneous determination of levetiracetam and lamotrigine in whole blood.
Journal of pharmaceutical and biomedical analysis (2014-09-23)
Panagiota Nikolaou, Ioannis Papoutsis, Artemisia Dona, Chara Spiliopoulou, Sotiris Athanaselis
ABSTRACT
A sensitive and accurate gas chromatography-mass spectrometric method was developed and validated for the simultaneous determination of levetiracetam and lamotrigine in whole blood. A solid-phase extraction (SPE) procedure using HF Bond Elut C18 columns followed by derivatization using N-methyl-N-tert-butyldimethylsilyl-trifluoroacetamide (MTBSTFA) with 1% tert-butyldimethylsilyl chloride (TBDMSCl) was used. In this assay, levetiracetam-d6 was used as internal standard. Limits of detection and quantification were 0.15 and 0.50 μg/mL, respectively, for both analytes. The method was proved to be linear within the concentration range of 0.50-50.0 μg/mL (R(2) ≥ 0.992) for both analytes. Absolute recovery was found to be at least 90.0 and 97.2% for levetiracetam and lamotrigine, respectively. Intra-day and inter-day accuracy values for both analytes were ranged from -6.5 to 4.2 and -6.6 to 3.0%, respectively, whereas their respective precision values were less than 11.4 and 8.3%. The developed method was successfully used in our laboratory for quantification of levetiracetam and lamotrigine blood concentrations during the investigation of forensic cases where these antiepileptic drugs were involved. This method could also be used for therapeutic drug monitoring purposes.
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Supelco
Heptafluorobutyric anhydride, for GC derivatization, LiChropur, ≥99.0%
Sigma-Aldrich
N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide with 1% tert-Butyldimethylchlorosilane, ≥95%
Sigma-Aldrich
Chlorotrimethylsilane, purified by redistillation, ≥99%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Supelco
Heptafluorobutyric anhydride, for GC derivatization, LiChropur, ≥99.0%
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, for GC derivatization, LiChropur, contains 10% TMCS, 98% (excluding TMCS)
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, for GC derivatization, LiChropur, contains 1% TMCS, 99% (excluding TMCS)
Sigma-Aldrich
tert-Butylchlorodimethylsilane solution, 50 wt. % in toluene
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Chlorotrimethylsilane, ≥98.0% (GC)
Sigma-Aldrich
Chlorotrimethylsilane, puriss., ≥99.0% (GC)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Heptafluorobutyric anhydride, ≥98%
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, with 1% trimethylchlorosilane, for GC derivatization, LiChropur
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Trifluoroacetic anhydride, for GC derivatization, LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Chlorotrimethylsilane, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Hydrofluoric acid, ACS reagent, 48%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%