MilliporeSigma
  • Conditioned medium of adipose-derived stromal cell culture in three-dimensional bioreactors for enhanced wound healing.
Conditioned medium of adipose-derived stromal cell culture in three-dimensional bioreactors for enhanced wound healing.
The Journal of surgical research (2014-12-06)
Sun Hyun Kwon, Suk Ho Bhang, Hyeon-Ki Jang, Taiyoun Rhim, Byung-Soo Kim
ABSTRACT
It was previously shown that human adipose-derived stromal cell (hADSC)-conditioned medium (CM) promotes wound healing. An essential part of the wound healing process is neovascularization in the wound bed. We hypothesized that CM prepared from hADSCs cultured as spheroids in three-dimensional suspension bioreactors (spheroid CM) would contain much higher concentrations of angiogenic growth factors secreted by hADSCs, induce a higher extent of neovascularization in the wound bed, and improve wound healing as compared with CM prepared by conventional monolayer culture (monolayer CM). The concentrations of angiogenic growth factors (i.e., vascular endothelial growth factor, basic fibroblast growth factor, and hepatocyte growth factor) in spheroid CM were 20- to 145-fold higher than those in monolayer CM. Either fresh medium, monolayer CM, or spheroid CM was administered to full-thickness wounds created on the dorsal aspects of athymic mice. The monolayer CM promoted wound healing as compared with fresh medium or no treatment. Importantly, wound closure was faster, and dermal and epidermal regeneration was improved in the spheroid CM-treated mice compared with that in the monolayer CM-treated mice. The improved wound healing by spheroid CM may be attributed, at least in part, to enhanced neovascularization in the wound beds. The spheroid-based CM approach showed potential as a therapy for skin wound repair.
Sigma-Aldrich
Chloroform, suitable for HPLC
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, SAJ super special grade, ≥99.0%
Sigma-Aldrich
Chloroform, SAJ first grade, ≥99.0%, contains 0.4-0.8% ethanol
Sigma-Aldrich
Chloroform, JIS 300, ≥99.0%, for residue analysis
Sigma-Aldrich
Chloroform, JIS special grade, ≥99.0%
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, amylene stabilized
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)