MilliporeSigma
  • Multi-analyte method for the analysis of various organohalogen compounds in house dust.
Multi-analyte method for the analysis of various organohalogen compounds in house dust.
Analytica chimica acta (2014-12-07)
Darina Lankova, Andrea Svarcova, Kamila Kalachova, Ondrej Lacina, Jana Pulkrabova, Jana Hajslova
ABSTRACT
In the present study, a novel analytical approach for the simultaneous determination of 27 brominated flame retardants (BFRs), namely polybrominated diphenyl ethers (PBDEs), isomers of hexabromocyclododecane (HBCD), tetrabromobisphenol A (TBBPA) and several novel BFRs (NBFRs), together with 18 perfluoroalkyl substances (PFASs) in indoor dust was developed and validated. To achieve integrated isolation of analytes from the sample and their fractionation, a miniaturized method based on matrix solid phase dispersion (MSPD) was employed. Principally, after mixing the dust (<0.1 g) with the Florisil(®), the mixture was applied on the top of a sorbent (Florisil(®)) placed in glass column and then analytes were eluted using solvents with different polarities. For the identification/quantification of target compounds largely differing in polarity, complementary techniques represented by gas and liquid chromatography coupled to tandem mass spectrometry (GC-MS/MS and LC-MS/MS) were used. The results of validation experiments, which were performed on the SRM 2585 material (for PBDEs, HBCDs and TBBPA), were in accordance with the certified/reference values. For other analytes (NBFRs and PFASs), the analysis of an artificially contaminated blank dust sample was realized. The method recoveries for all target compounds ranged from 81 to 122% with relative standard deviations lower than 21%. The quantification limits were in the range of 1-25 ng g(-1) for BFRs and 0.25-1 ng g(-1) for PFASs. Finally, 18 samples (6 households × 3 sampling sites) were analyzed. The high variability between concentrations of PFASs and BFRs in the dust samples from various households as well as collecting sites in a respective house was observed. The total amounts of PFASs and BFRs were in the range of 1.58-236 ng g(-1) (median 10.6 ng g(-1)) and 39.2-2320 ng g(-1) (median 325 ng g(-1)), respectively. It was clearly shown that dust from the indoor environment might be a significant source of human exposure to various organohalogen pollutants.
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Supelco
Acetone, analytical standard
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Acetone, suitable for HPLC
Sigma-Aldrich
Acetone, for chromatography, ≥99.8%
Sigma-Aldrich
Acetone, for residue analysis, ≥99.5%
Sigma-Aldrich
Ammonium acetate solution, 50 % (w/v), for copper determination
Sigma-Aldrich
Acetone, ≥99.5%, for residue analysis
Sigma-Aldrich
Acetone, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetone, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetone, for residue analysis, JIS 5000
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Supelco
Density Standard 692 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)