MilliporeSigma
  • Androctonus australis hector venom contributes to the interaction between neuropeptides and mast cells in pulmonary hyperresponsiveness.
Androctonus australis hector venom contributes to the interaction between neuropeptides and mast cells in pulmonary hyperresponsiveness.
International immunopharmacology (2015-01-21)
Imène Chaïr-Yousfi, Fatima Laraba-Djebari, Djelila Hammoudi-Triki
ABSTRACT
Lung injury and respiratory distress syndrome are frequent symptoms observed in the most severe cases of scorpion envenomation. The uncontrolled transmigration of leukocyte cells into the lung interstitium and alveolar space and pulmonary edema may be the cause of death. Mast cells can release various inflammatory mediators known to be involved in the development of lung edema following scorpion venom injection. The present study was designed to determine the evidence of neurokinin 1 (NK1) receptor and the involvement of mast cell activation to induce pulmonary edema and to increase vascular permeability after Androctonus australis hector (Aah) venom administration. To this end, mast cells were depleted using compound 48/80 (C48/80). Furthermore, the involvement of tachykinin NK1 receptors expressed on mast cell membranes was elucidated by their blocking with an antagonist. On the other hand, the ability of Aah venom to increase vascular permeability and to induce edema was also assessed by measuring the amount of Evans blue dye (EBD) extravasation in bronchoalveolar lavage (BAL) fluid and in the lungs of mice. Pulmonary edema, as assessed by the levels of EBD extravasation, was completely inhibited in compound 48/80-treated animals. Depletion by stimuli non-immunological C48/80 component markedly reduced induced inflammatory response following the venom administration. The mast cells seem to play an important role in the development of lung injury and the increase of vascular permeability in mice following the subcutaneous administration of Aah scorpion venom through the NK1 receptor.
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Pricing and availability is not currently available.
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
o-Dianisidine dihydrochloride, Suitable for use in glucose determination
Pricing and availability is not currently available.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Pricing and availability is not currently available.
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Pricing and availability is not currently available.
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Pricing and availability is not currently available.
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrochloric acid solution, 0.5 M
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrochloric acid solution, 0.05 M
Pricing and availability is not currently available.
Sigma-Aldrich
N,N-Dimethylformamide, SAJ first grade, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfuric acid solution, 0.01 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfuric acid, JIS special grade, ≥95.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrochloric acid solution, 1 M
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrochloric acid solution, 12 M
Pricing and availability is not currently available.
Sigma-Aldrich
Acetic acid solution, 1 M, 1 N
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfuric acid solution, 0.25 M
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrochloric acid, JIS special grade, 35.0-37.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrochloric acid solution, 0.2 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfuric acid solution, 0.5 M
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen chloride – ethanol solution, 0.1 M in ethanol
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrochloric acid solution, 2 M
Pricing and availability is not currently available.
Sigma-Aldrich
Acetic acid, ≥99.7%
Pricing and availability is not currently available.