MilliporeSigma
  • Activated-ester-type photocleavable crosslinker for preparation of photodegradable hydrogels using a two-component mixing reaction.
Activated-ester-type photocleavable crosslinker for preparation of photodegradable hydrogels using a two-component mixing reaction.
Advanced healthcare materials (2014-08-15)
Fumiki Yanagawa, Shinji Sugiura, Toshiyuki Takagi, Kimio Sumaru, Gulden Camci-Unal, Alpesh Patel, Ali Khademhosseini, Toshiyuki Kanamori
ABSTRACT
Photodegradable hydrogels have emerged as powerful platforms for studying and directing cellular behavior in a spatiotemporally controlled manner. Photodegradable hydrogels have previously been formed by free radical polymerizations, Michael-type addition reactions, and orthogonal click reactions. Here, an ester-activated photocleavable crosslinker is presented for preparing photodegradable hydrogels by means of a one-step mixing reaction between the crosslinker and a biocompatible polymer containing amino moieties (amino-terminated tetra-arm poly(ethylene glycol) or gelatin). It is demonstrated that photodegradable hydrogels micropatterned by photolithography can be used to culture cells with high viability and proliferation rates. The resulting micropatterned cell-laden structures can potentially be used to create 3D biomaterials for various tissue-engineering applications.
Sigma-Aldrich
N-Hydroxysuccinimide, purum, ≥97.0% (T)
Sigma-Aldrich
Butyric acid, SAJ special grade, ≥99.5%
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, commercial grade, powder
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, crystalline
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, ≥99.0% (AT)
Supelco
Butyric acid, analytical standard
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, purum, ≥98.0% (AT)
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
Butyric acid, ≥99%, FG
Sigma-Aldrich
Butyric acid, ≥99%
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, BioXtra
Sigma-Aldrich
Butyric acid, natural, ≥99%, FCC, FG
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet