MilliporeSigma
  • High-performance liquid chromatographic enantioseparation of unusual amino acid derivatives with axial chirality on polysaccharide-based chiral stationary phases.
High-performance liquid chromatographic enantioseparation of unusual amino acid derivatives with axial chirality on polysaccharide-based chiral stationary phases.
Journal of chromatography. A (2015-03-10)
Pilar López-Ram-de-Víu, José A Gálvez, María D Díaz-de-Villegas
ABSTRACT
The successful enantioseparation of axially chiral amino acid derivatives containing a cyclohexylidene moiety on an analytical and semipreparative scale was achieved for the first time by HPLC using polysaccharide-based chiral stationary phases. Racemic methyl N-benzoylamino esters, easily obtained by methanolysis of the corresponding 5(4H)-oxazolones, were subjected to chiral HPLC resolution using chiral stationary phases based on immobilized 3,5-dimethylphenylcarbamate derivatives of amylose (Chiralpak(®) IA column) or cellulose (Chiralpak(®) IB column). The behaviour of both selectors under different elution conditions was evaluated and compared. The amylose column showed better performance than the cellulose column for all enantiomers tested. The semipreparative resolution of axially chiral amino acid derivatives with different side chains has been achieved on a 250mm×20mm ID Chiralpak(®) IA column using the appropriate mixture of n-hexane/chlorofom/ethanol as eluent by successive injections of a solution of the sample in chloroform. Using this protocol up to 120mg of each enantiomer of the corresponding axially chiral amino acid derivative were obtained from 300mg of racemate. [(Sa)-2a, 105mg; (Ra)-2a, 60mg, [(Sa)-2b, 105mg; (Ra)-2b, 90mg, [(Sa)-2c, 120mg; (Ra)-2c, 100mg].
Product Description
Supelco
Chloroform, analytical standard
Supelco
Acetone, analytical standard
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Chloroform, suitable for HPLC
Sigma-Aldrich
Acetone, suitable for HPLC
Sigma-Aldrich
Acetone, for chromatography, ≥99.8%
Sigma-Aldrich
Chloroform, SAJ first grade, ≥99.0%, contains 0.4-0.8% ethanol
Sigma-Aldrich
Acetone, for residue analysis, ≥99.5%
Sigma-Aldrich
Chloroform, JIS 300, ≥99.0%, for residue analysis
Sigma-Aldrich
Chloroform, JIS special grade, ≥99.0%
Sigma-Aldrich
Acetone, ≥99.5%, for residue analysis
Sigma-Aldrich
Acetone, JIS special grade, ≥99.5%
Sigma-Aldrich
Chloroform, SAJ super special grade, ≥99.0%
Sigma-Aldrich
Acetone, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetone, for residue analysis, JIS 5000
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer