MilliporeSigma
  • Increased antidepressant sensitivity after prefrontal cortex glucocorticoid receptor gene deletion in mice.
Increased antidepressant sensitivity after prefrontal cortex glucocorticoid receptor gene deletion in mice.
Physiology & behavior (2014-12-03)
Rifat J Hussain, Lauren Jacobson
ABSTRACT
Our laboratory has previously shown that antidepressants regulate glucocorticoid receptor (GR) expression in the prefrontal cortex (PFC). To determine if PFC GR are involved in antidepressant effects on behavior or hypothalamic-pituitary-adrenocortical (HPA) axis activity, we treated floxed GR male mice with saline or 15 or 30 mg/kg/d imipramine after PFC injection of adeno-associated virus 2/9 vectors transducing expression of Cre recombinase, to knock-down GR (PFC-GRKD), or green fluorescent protein (PFC-GFP), to serve as a control. The pattern of virally transduced GR deletion, common to all imipramine treatment groups, included the infralimbic, prelimbic, and medial anterior cingulate cortex at its largest extent, but was confined to the prelimbic and anterior cingulate cortex at its smallest extent. PFC GR knock-down increased behavioral sensitivity to imipramine, with imipramine-treated PFC-GRKD but not PFC-GFP mice exhibiting significant decreases in depression-like immobility during forced swim. PFC GR deletion did not alter general locomotor activity. The 30 mg/kg dose of imipramine increased plasma corticosterone levels immediately after a 5-min forced swim, but PFC GR knock-down had no significant effect on plasma corticosterone under these experimental conditions. We conclude that PFC GR knock-down, likely limited to the medial prelimbic and anterior cingulate cortices, can increase behavioral sensitivity to antidepressants. These findings indicate a novel role for PFC GR in influencing antidepressant response.
Product Description
Supelco
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethylene glycol, JIS special grade, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethylene glycol, SAJ first grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Ethylene glycol solution, NMR reference standard, 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
SKU
Pack Size
Availability
Price
Quantity
USP
Ethylene glycol, United States Pharmacopeia (USP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Indomethacin, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Corticosterone, ≥92%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Indomethacin, meets USP testing specifications
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Corticosterone, ≥98.5% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
SKU
Pack Size
Availability
Price
Quantity
Supelco
Corticosterone, VETRANAL®, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Supelco
Ethylene glycol, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethylene glycol, ReagentPlus®, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethylene glycol, spectrophotometric grade, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
SKU
Pack Size
Availability
Price
Quantity
USP
Indomethacin, United States Pharmacopeia (USP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Supelco
Indomethacin, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
SKU
Pack Size
Availability
Price
Quantity
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycerol, FCC, FG
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)
SKU
Pack Size
Availability
Price
Quantity
Supelco
Glycerol, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethylene glycol 5 M solution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycerol, JIS special grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycerol, SAJ first grade, ≥98.0%