MilliporeSigma
  • Collecting duct-specific knockout of renin attenuates angiotensin II-induced hypertension.
Collecting duct-specific knockout of renin attenuates angiotensin II-induced hypertension.
American journal of physiology. Renal physiology (2014-08-15)
Nirupama Ramkumar, Deborah Stuart, Sara Rees, Alfred Van Hoek, Curt D Sigmund, Donald E Kohan
ABSTRACT
The physiological and pathophysiological significance of collecting duct (CD)-derived renin, particularly as it relates to blood pressure (BP) regulation, is unknown. To address this question, we generated CD-specific renin knockout (KO) mice and examined BP and renal salt and water excretion. Mice containing loxP-flanked exon 1 of the renin gene were crossed with mice transgenic for aquaporin-2-Cre recombinase to achieve CD-specific renin KO. Compared with controls, CD renin KO mice had 70% lower medullary renin mRNA and 90% lower renin mRNA in microdissected cortical CD. Urinary renin levels were significantly lower in KO mice (45% of control levels) while plasma renin concentration was significantly higher in KO mice (63% higher than controls) during normal-Na intake. While no observable differences were noted in BP between the two groups with varying Na intake, infusion of angiotensin II at 400 ng·kg(-1)·min(-1) resulted in an attenuated hypertensive response in the KO mice (mean arterial pressure 111 ± 4 mmHg in KO vs. 128 ± 3 mmHg in controls). Urinary renin excretion and epithelial Na(+) channel (ENaC) remained significantly lower in the KO mice following ANG II infusion compared with controls. Furthermore, membrane-associated ENaC protein levels were significantly lower in KO mice following ANG II infusion. These findings suggest that CD renin modulates BP in ANG II-infused hypertension and these effects are associated with changes in ENaC expression.
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Pricing and availability is not currently available.
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Pricing and availability is not currently available.
Trolamine, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
SAFC
BIS-TRIS
Pricing and availability is not currently available.
SAFC
BIS-TRIS
Pricing and availability is not currently available.
Sigma-Aldrich
Sucrose, ACS reagent
Pricing and availability is not currently available.
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Pricing and availability is not currently available.
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Pricing and availability is not currently available.
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Triethanolamine, BioUltra, ≥99.5% (GC)
Pricing and availability is not currently available.
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Triethanolamine, puriss. p.a., ≥99% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sucrose, meets USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Pricing and availability is not currently available.
Sucrose, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
Triethanolamine, ≥99.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sucrose, SAJ first grade
Pricing and availability is not currently available.
Sigma-Aldrich
Triethanolamine, JIS special grade, ≥98.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sucrose, JIS special grade
Pricing and availability is not currently available.
Sigma-Aldrich
Triethanolamine, reagent grade, 98%
Pricing and availability is not currently available.
Supelco
Triethanolamine, analytical standard
Pricing and availability is not currently available.