MilliporeSigma
  • Occurrence, removal, and fate of progestogens, androgens, estrogens, and phenols in six sewage treatment plants around Dianchi Lake in China.
Occurrence, removal, and fate of progestogens, androgens, estrogens, and phenols in six sewage treatment plants around Dianchi Lake in China.
Environmental science and pollution research international (2014-07-01)
Bin Huang, Xiaoman Li, Wenwen Sun, Dong Ren, Xiao Li, Xiaonan Li, Ying Liu, Qiang Li, Xuejun Pan
ABSTRACT
The occurrence and behavior of endocrine disrupting chemicals (EDCs) in sewage treatment plants (STPs), especially estrogens and phenols, have been closely concerned in previous studies. However, the systematical researches about progestogens and androgens were scarce in STPs adopting different treatment technologies. This work investigated the occurrence, removal, and fate of one progestogen, three androgens, four estrogens, and six phenols in six STPs around Dianchi Lake in China, where the influents, effluents of primary treatment, secondary treatment, and advanced treatment, as well as excess sludge samples, were analyzed. All of the above EDCs were detected out in influents of the six STPs. Bisphenol A, nonylphenol-mono-ethoxylate, and nonylphenol-diethoxylate were the dominant EDCs detected in those influent samples with the concentrations that varied from 637.6 to 1,684.0 ng/L, 633.8 to 1,540.0 ng/L, and 648.7 to 2,246.0 ng/L, respectively; E1 and dihydrotestosterone were the major steroids with the mean concentration of 126.8 and 277.4 ng/L. For effluents and sludges, phenols showed higher concentration (366.8-1,233.0 ng/L and 1,478.1-6,948.9 ng/g dry weight (dw)) and detection rate (100 %). The total removal rates were more than 80 % for most compounds in wastewater treatment processes, and high removal efficiency (86-100 %) was found for androgens and progestogens compared with estrogens (75-92 %) and phenols (62-85 %). The secondary treatment processes play significant roles on degrading EDCs, whereas the primary sedimentation has little effects. The treatment capacity of anoxic-anaerobic-anoxic membrane bioreactor and anaerobic/anoxic/oxic technologies was superior to the conventional oxidation ditch in the degradation of EDCs. The advanced treatment process, two units of filter (D-type or V-type), and ultraviolet disinfection were adopted and presented effective to remove these compounds. According to fate analysis, it was obvious that biological degradation was the main pathway on the removal of EDCs in STPs compared with adsorption. Risk quotients were calculated to assess ecological risks of those EDCs. Risk quotients of 54 and 61 % were more than 1 in effluents and sludges, respectively, showing potential hazard of effluents and sludges to the environment.
Sigma-Aldrich
Cyclohexane, JIS special grade
Sigma-Aldrich
Cyclohexane, suitable for HPLC
Supelco
4-tert-Octylphenol, analytical standard
Sigma-Aldrich
4-tert-Octylphenol, 97%
Sigma-Aldrich
Estrone, ≥99%
Sigma-Aldrich
Cyclohexane, SAJ first grade, ≥99.0%
USP
Estrone, United States Pharmacopeia (USP) Reference Standard
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, for GC derivatization, LiChropur, contains 10% TMCS, 98% (excluding TMCS)
Supelco
N-Methyl-bis(trifluoroacetamide), for GC derivatization, LiChropur, ≥97.0% (GC)
Sigma-Aldrich
5α-Androstane
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, with 1% trimethylchlorosilane, for GC derivatization, LiChropur
Sigma-Aldrich
Testosterone, ≥98%
Sigma-Aldrich
Chlorotrimethylsilane, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, for GC derivatization, LiChropur, contains 1% TMCS, 99% (excluding TMCS)
Sigma-Aldrich
Chlorotrimethylsilane, ≥98.0% (GC)
Supelco
Testosterone, VETRANAL®, analytical standard
Supelco
N-Methyl-bis(trifluoroacetamide), for GC derivatization, LiChropur, ≥99.0% (GC)
Supelco
Estrone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cyclohexane, ACS reagent, ≥99%
Sigma-Aldrich
Cyclohexane, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Cyclohexane, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Cyclohexane, Laboratory Reagent, ≥99.8%
Sigma-Aldrich
Cyclohexane, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
Cyclohexane, ACS reagent, ≥99%
Supelco
Chlorotrimethylsilane, for GC derivatization, LiChropur, ≥99.0% (GC)
Testosterone, European Pharmacopoeia (EP) Reference Standard
Supelco
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Estrone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Chlorotrimethylsilane, puriss., ≥99.0% (GC)
Sigma-Aldrich
Iodotrimethylsilane, 97%