MilliporeSigma
  • Preparation, characteristics and assessment of a novel gelatin-chitosan sponge scaffold as skin tissue engineering material.
Preparation, characteristics and assessment of a novel gelatin-chitosan sponge scaffold as skin tissue engineering material.
International journal of pharmaceutics (2014-10-03)
Fei Han, Yang Dong, Zhen Su, Ran Yin, Aihua Song, Sanming Li
ABSTRACT
In order to develop a skin tissue engineering material for wound dressing application, a novel gelatin-chitosan sponge scaffold was designed and studied. The effect of chitosan and gelatin ratio on the morphology, pore size, porosity, water uptake capacity, water retention capacity and the degradation behavior were evaluated. Biocompatibility was investigated by both MTT method and AO/EB staining method. Antibacterial assessment and in vivo pharmacodynamic was also studied to evaluate the potential for wound healing. Results showed the sponge scaffold have uniform porous structure with pore size range between 120 and 140 μm, high porosity (>90%), high water uptake capacity (>1500%), high water retention capacity (>400%), and degradation percent in 28 days between 38.3 and 53.9%. Biocompatibility results showed that the activity of cells could not be affected by the nature of the sponge and it was suitable for cell adhesion and proliferation for 21 days. In vivo evaluation indicated that the sponge scaffold could offer effective support and attachment to cells for skin wound healing. In conclusion, the developed sponge scaffold was a potential skin tissue engineering material with appropriate physical properties and good biocompatibility.
Product Description
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethanol, ≥99.5%
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for fluorescence
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, for residue analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, for residue analysis
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, puriss. p.a., absolute, ≥99.8% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%