Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, AnhydroBeads™, −10 mesh, 99.999% trace metals basis
Pricing and availability is not currently available.
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, tablet
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 5 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 1 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Pricing and availability is not currently available.
Paclitaxel semi-synthetic for peak identification, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Trolamine, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Glutathione, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
SAFC
Sodium chloride solution, 5 M
Pricing and availability is not currently available.
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.