MilliporeSigma
Unsaturated fatty acids induce non-canonical autophagy.
The EMBO journal (2015-01-15)
Mireia Niso-Santano, Shoaib Ahmad Malik, Federico Pietrocola, José Manuel Bravo-San Pedro, Guillermo Mariño, Valentina Cianfanelli, Amena Ben-Younès, Rodrigo Troncoso, Maria Markaki, Valentina Sica, Valentina Izzo, Kariman Chaba, Chantal Bauvy, Nicolas Dupont, Oliver Kepp, Patrick Rockenfeller, Heimo Wolinski, Frank Madeo, Sergio Lavandero, Patrice Codogno, Francis Harper, Gérard Pierron, Nektarios Tavernarakis, Francesco Cecconi, Maria Chiara Maiuri, Lorenzo Galluzzi, Guido Kroemer
ABSTRACT
To obtain mechanistic insights into the cross talk between lipolysis and autophagy, two key metabolic responses to starvation, we screened the autophagy-inducing potential of a panel of fatty acids in human cancer cells. Both saturated and unsaturated fatty acids such as palmitate and oleate, respectively, triggered autophagy, but the underlying molecular mechanisms differed. Oleate, but not palmitate, stimulated an autophagic response that required an intact Golgi apparatus. Conversely, autophagy triggered by palmitate, but not oleate, required AMPK, PKR and JNK1 and involved the activation of the BECN1/PIK3C3 lipid kinase complex. Accordingly, the downregulation of BECN1 and PIK3C3 abolished palmitate-induced, but not oleate-induced, autophagy in human cancer cells. Moreover, Becn1(+/-) mice as well as yeast cells and nematodes lacking the ortholog of human BECN1 mounted an autophagic response to oleate, but not palmitate. Thus, unsaturated fatty acids induce a non-canonical, phylogenetically conserved, autophagic response that in mammalian cells relies on the Golgi apparatus.
Sigma-Aldrich
Monoclonal Anti-ATG5 antibody produced in mouse, clone ATG5-18, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-ATG7 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution