MilliporeSigma
  • High-mobility group box 1: a novel target for treatment of Pseudomonas aeruginosa keratitis.
High-mobility group box 1: a novel target for treatment of Pseudomonas aeruginosa keratitis.
Journal of immunology (Baltimore, Md. : 1950) (2015-01-16)
Sharon McClellan, Xiaoyu Jiang, Ronald Barrett, Linda D Hazlett
ABSTRACT
High-mobility group box 1 (HMGB1), a prototypic alarmin, mediates the systemic inflammatory response syndrome. Treatment with vasoactive intestinal peptide, an anti-inflammatory neuropeptide, downregulates proinflammatory cytokines and promotes healing in a susceptible (cornea perforates) model of Pseudomonas aeruginosa keratitis, and also significantly downregulates HMGB1 expression. Therefore, we examined targeting HMGB1 for the treatment of P. aeruginosa keratitis to avoid delivery and other issues associated with vasoactive intestinal peptide. For this, HMGB1 was silenced using small interfering RNA, whereas controls were treated with a nonspecific scrambled sequence small interfering RNA. Less disease was seen postinfection in siHMGB1 compared with control mice and was documented by clinical score and photographs with a slit lamp. Real-time RT-PCR and ELISA confirmed HMGB1 knockdown. RT-PCR analysis also revealed reduced mRNA levels of IL-1β, MIP-2, TNF-α, TLR4, and receptor for advanced glycation end products, whereas mRNA levels of anti-inflammatory TLRs single Ig IL-1-related receptor and ST2 were increased significantly. HMGB1 knockdown also decreased IL-1β and MIP-2 proteins, reducing polymorphonuclear cell number in the infected cornea. mRNA and protein levels of CXCL12 and CXCR4, as well as mononuclear cells, were reduced significantly after HMGB1 knockdown. Ab neutralization of HMGB1, infection with a clinical isolate, and recombinant HMGB1 treatment of resistant mice supported the silencing studies. These data provide evidence that silencing HMGB1 promotes better resolution of P. aeruginosa keratitis by decreasing levels of proinflammatory mediators (decreasing polymorphonuclear cell infiltration), increasing anti-inflammatory TLRs, reducing CXCL12 (preventing HMGB1/CXCL12 heterodimer formation), and signaling through CXCR4, reducing monocyte/macrophage infiltration.
Sigma-Aldrich
Sodium trichloroacetate, 97%
Pricing and availability is not currently available.
Supelco
Chloroform, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
o-Dianisidine dihydrochloride, Suitable for use in glucose determination
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Pricing and availability is not currently available.
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Pricing and availability is not currently available.
Sigma-Aldrich
Vasoactive Intestinal Peptide human, porcine, rat, ≥95% (HPLC), powder
Pricing and availability is not currently available.
Sigma-Aldrich
Vasoactive Intestinal Peptide human, porcine, rat, synthetic, ≥95% (HPLC), powder
Pricing and availability is not currently available.
Sigma-Aldrich
o-Dianisidine dihydrochloride, ≥95%
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Pricing and availability is not currently available.
Sigma-Aldrich
o-Dianisidine dihydrochloride, tablet, 10 mg substrate per tablet
Pricing and availability is not currently available.
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform, suitable for HPLC
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform, SAJ first grade, ≥99.0%, contains 0.4-0.8% ethanol
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform, JIS 300, ≥99.0%, for residue analysis
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform, JIS special grade, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform, SAJ super special grade, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Pricing and availability is not currently available.
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Pricing and availability is not currently available.