MilliporeSigma
  • Temporal changes in glutaredoxin 1 and protein s-glutathionylation in allergic airway inflammation.
Temporal changes in glutaredoxin 1 and protein s-glutathionylation in allergic airway inflammation.
PloS one (2015-04-16)
Kanako Maki, Katsura Nagai, Masaru Suzuki, Takashi Inomata, Takayuki Yoshida, Masaharu Nishimura
ABSTRACT
Asthma is a chronic inflammatory disorder of the airways, involving oxidative stress. Upon oxidative stress, glutathione covalently binds to protein thiols to protect them against irreversible oxidation. This posttranslational modification, known as protein S-glutathionylation, can be reversed by glutaredoxin 1 (Glrx1) under physiological condition. Glrx1 is known to increase in the lung tissues of a murine model of allergic airway inflammation. However, the temporal relationship between levels of Glrx1, protein S-glutathionylation, and glutathione in the lungs with allergic airway inflammation is not clearly understood. BALB/c mice received 3 aerosol challenges with ovalbumin (OVA) following sensitization to OVA. They were sacrificed at 6, 24, 48, or 72 h, or 8 days (5 mice per group), and the levels of Glrx1, protein S-glutathionylation, glutathione, and 25 cytokines/chemokines were evaluated in bronchoalveolar lavage fluid (BALF) and/or lung tissue. Levels of Glrx1 in BALF were significantly elevated in the OVA 6 h (final challenge) group compared to those in the control, with concurrent increases in protein S-glutathionylation levels in the lungs, as well as total glutathione (reduced and oxidized) and oxidized glutathione in BALF. Protein S-glutathionylation levels were attenuated at 24 h, with significant increases in Glrx1 levels in lung tissues at 48 and 72 h. Glrx1 in alveolar macrophages was induced after 6 h. Glrx1 levels concomitantly increased with Th2/NF-κB-related cytokines and chemokines in BALF. The temporal relationships of Glrx1 with protein S-glutathionylation, glutathione, and cytokines/chemokines were observed as dynamic changes in lungs with allergic airway inflammation, suggesting that Glrx1 and protein-SSG redox status may play important roles in the development of allergic airway inflammation.
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Pricing and availability is not currently available.
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, BioReagent, suitable for insect cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Neocuproine, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
Biocytin, ≥98% (TLC)
Pricing and availability is not currently available.
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Pricing and availability is not currently available.
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, BioXtra, ≥98.0%
Pricing and availability is not currently available.
Supelco
Meprobamate solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.
Trolamine, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Glutathione, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
Aluminum hydroxide, reagent grade
Pricing and availability is not currently available.
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, ACS reagent, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
L-Glutathione oxidized disodium salt, ≥98%, powder
Pricing and availability is not currently available.
Sigma-Aldrich
L-Glutathione oxidized disodium salt, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Pricing and availability is not currently available.
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Pricing and availability is not currently available.