MilliporeSigma
  • Dissolution enhancement and in vitro performance of clarithromycin nanocrystals produced by precipitation-lyophilization-homogenization method.
Dissolution enhancement and in vitro performance of clarithromycin nanocrystals produced by precipitation-lyophilization-homogenization method.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2014-09-10)
Boontida Morakul, Jiraphong Suksiriworapong, Mullika Traidej Chomnawang, Peter Langguth, Varaporn Burapacheep Junyaprasert
ABSTRACT
The gastroduodenal diseases caused by Helicobacter pylori were commonly treated with antibiotic clarithromycin as a standard regimen. According to the poorly water-soluble of clarithromycin, the nanocrystal formulation was prepared. The aim of this study was to investigate an enhancement effect of clarithromycin nanocrystals produced by precipitation-lyophilization-homogenization (PLH) method on the saturation solubility, dissolution velocity, antibiotic activity, permeability through the gastric mucus and cellular permeability. Poloxamer 407 and sodium lauryl sulfate (SLS) were chosen as combined stabilizers in the nanocrystal system. The obtained clarithromycin nanocrystals were identified as cubic particles by SEM with a bulk population of approximately 400nm existed in crystalline and/or partial amorphous form as investigated by DSC and XRPD. The saturation solubility of the clarithromycin nanocrystals was increased by 1.5- and 6-folds higher than clarithromycin powder in buffer pH 5.0 and 6.8, respectively. The dissolution profiles of clarithromycin nanocrystals at pH 5.0 and 6.8 were significantly different from clarithromycin powder and the marketed product (f1 value >15 and f2 value <50). All dissolution parameters (relative dissolution rate, percent dissolution efficiency and mean dissolution time) showed that clarithromycin nanocrystals had higher dissolution rate when compared with the clarithromycin powder, the lyophilized coarse suspension and the marketed product. The bioassay study by diffusion agar method showed a maintained antibiotic activity of clarithromycin nanocrystals solubilized in buffer solution which was greater potency than the lyophilized coarse suspension and the clarithromycin powder. Additionally, the nanocrystals possessed higher permeability through gastric mucus and cellular monolayer of Caco-2 and NCI-N87 cells as compared to the lyophilized coarse suspension and the clarithromycin powder. The results indicated that, the developed clarithromycin nanocrystals were a potential delivery system that exerts more effectiveness in H. pylori eradication.
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
N,N′-Disuccinimidyl carbonate, purum, ≥95.0% (NMR)
Sigma-Aldrich
N,N′-Disuccinimidyl carbonate, ≥95%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Supelco
Clarithromycin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Clarithromycin, ≥95% (HPLC)
Sigma-Aldrich
Clarithromycin, 96.0-102.0% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Clarithromycin, European Pharmacopoeia (EP) Reference Standard
USP
Clarithromycin, United States Pharmacopeia (USP) Reference Standard
Clarithromycin for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium acetate trihydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate trihydrate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E262, 99.0-101.0% (calc. to the dried substance), ≤0.00002% Al