MilliporeSigma
  • Preventive effects of cranberry products on experimental colitis induced by dextran sulphate sodium in mice.
Preventive effects of cranberry products on experimental colitis induced by dextran sulphate sodium in mice.
Food chemistry (2014-08-26)
Xiao Xiao, Jonggun Kim, Quancai Sun, Daeyoung Kim, Cheon-Seok Park, Tzong-Shi Lu, Yeonhwa Park
ABSTRACT
With the prevalence of inflammatory bowel disease (IBD) and its associated risk for development of colorectal cancer, it is of great importance to prevent and treat IBD. However, due to the complexity of etiology and potentially serious adverse effects, treatment options for IBD are relatively limited. Thus, the purpose of this study was to identify a safe food-based approach for the prevention and treatment of IBD. In this study, we tested the effects of cranberry products on preventing dextran sulphate sodium-induced murine colitis. Our results suggest that both cranberry extract and dried cranberries-fed groups had a significantly reduced disease activity index, where dried cranberries were more effective in preventing colitis than cranberry extract. Shortening of colon length, colonic myeloperoxidase activity and production of pro-inflammatory cytokines were attenuated in animals fed dried cranberries compared to the controls. The current report suggests that cranberries can be applied to prevent and reduce the symptoms of IBD.
Sigma-Aldrich
o-Dianisidine dihydrochloride, Suitable for use in glucose determination
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium sulfate, JIS special grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen peroxide solution, SAJ first grade, ≥30.0%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
SKU
Pack Size
Availability
Price
Quantity
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
SKU
Pack Size
Availability
Price
Quantity
USP
Cetrimonium bromide, United States Pharmacopeia (USP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
o-Dianisidine dihydrochloride, ≥95%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
o-Dianisidine dihydrochloride, tablet, 10 mg substrate per tablet
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
SKU
Pack Size
Availability
Price
Quantity
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium sulfate, puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99.0-100.5% (calc. to the dried substance)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized