MilliporeSigma
  • Comparative effects of the anti-platelet drugs, clopidogrel, ticlopidine, and cilostazol on aspirin-induced gastric bleeding and damage in rats.
Comparative effects of the anti-platelet drugs, clopidogrel, ticlopidine, and cilostazol on aspirin-induced gastric bleeding and damage in rats.
Life sciences (2014-07-02)
Koji Takeuchi, Shinichi Takayama, Chitose Izuhara
ABSTRACT
The present study compared the effects of frequently used anti-platelet drugs, such as clopidogrel, ticlopidine, and cilostazol, on the gastric bleeding and ulcerogenic responses induced by intraluminal perfusion with 25 mM aspirin acidified with 25 mM HCl (acidified ASA) in rats. The stomach was perfused with acidified ASA at a rate of 0.4 ml/min for 60 min under urethane anesthesia, and gastric bleeding was measured as the concentration of hemoglobin in the luminal perfusate, which was collected every 15 min. Clopidogrel (10-100mg/kg), ticlopidine (10-300 mg/kg), or cilostazol (3-30 mg/kg) was given p.o. 24h or 90 min before the perfusion of acidified ASA, respectively. Perfusion of the stomach with acidified ASA alone led to slight bleeding and lesions in the stomach. The pretreatment with clopidogrel, even though it did not cause bleeding or damage by itself, dose-dependently increased the gastric bleeding and ulcerogenic responses induced by acidified ASA. Ticlopidine also aggravated the severity of damage by increasing gastric bleeding, and the effects of ticlopidine at 300 mg/kg were equivalent to those of clopidogrel at 100mg/kg. In contrast, cilostazol dose-dependently decreased gastric bleeding and damage in response to acidified ASA. These results demonstrated that clopidogrel and ticlopidine, P2Y12 receptor inhibitors, increased gastric bleeding and ulcerogenic responses to acidified ASA, to the same extent, while cilostazol, a phosphodiesterase III inhibitor, suppressed these responses. Therefore, cilostazol may be safely used in dual anti-platelet therapy combined with ASA, without increasing the risk of gastric bleeding.
Sigma-Aldrich
o-Dianisidine dihydrochloride, Suitable for use in glucose determination
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
100 mL
In Stock
Details...
$51.90
6 x 20 mL
Estimated to ship on March 21, 2025
$60.60
Sigma-Aldrich
Cilostazol, ≥98% (HPLC), powder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
100 mL
In Stock
Details...
$51.90
6 x 20 mL
Estimated to ship on March 21, 2025
$60.60
USP
Cilostazol, United States Pharmacopeia (USP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
100 mL
In Stock
Details...
$51.90
6 x 20 mL
Estimated to ship on March 21, 2025
$60.60
USP
Cetrimonium bromide, United States Pharmacopeia (USP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
100 mL
In Stock
Details...
$51.90
6 x 20 mL
Estimated to ship on March 21, 2025
$60.60
Sigma-Aldrich
o-Dianisidine dihydrochloride, ≥95%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
100 mL
In Stock
Details...
$51.90
6 x 20 mL
Estimated to ship on March 21, 2025
$60.60
Sigma-Aldrich
o-Dianisidine dihydrochloride, tablet, 10 mg substrate per tablet
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
100 mL
In Stock
Details...
$51.90
6 x 20 mL
Estimated to ship on March 21, 2025
$60.60
Sigma-Aldrich
Urethane, ≥99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
100 mL
In Stock
Details...
$51.90
6 x 20 mL
Estimated to ship on March 21, 2025
$60.60
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥96.0% (AT)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
100 mL
In Stock
Details...
$51.90
6 x 20 mL
Estimated to ship on March 21, 2025
$60.60
Supelco
Hexadecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
100 mL
In Stock
Details...
$51.90
6 x 20 mL
Estimated to ship on March 21, 2025
$60.60
Supelco
Hexadecyltrimethylammonium bromide, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
100 mL
In Stock
Details...
$51.90
6 x 20 mL
Estimated to ship on March 21, 2025
$60.60
Sigma-Aldrich
Urethane, ≥99.0% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
100 mL
In Stock
Details...
$51.90
6 x 20 mL
Estimated to ship on March 21, 2025
$60.60
Supelco
Urethane, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
100 mL
In Stock
Details...
$51.90
6 x 20 mL
Estimated to ship on March 21, 2025
$60.60
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
100 mL
In Stock
Details...
$51.90
6 x 20 mL
Estimated to ship on March 21, 2025
$60.60
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
100 mL
In Stock
Details...
$51.90
6 x 20 mL
Estimated to ship on March 21, 2025
$60.60
SAFC
Hexadecyltrimethylammonium bromide, USP/NF
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
100 mL
In Stock
Details...
$51.90
6 x 20 mL
Estimated to ship on March 21, 2025
$60.60
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
100 mL
In Stock
Details...
$51.90
6 x 20 mL
Estimated to ship on March 21, 2025
$60.60
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
100 mL
In Stock
Details...
$51.90
6 x 20 mL
Estimated to ship on March 21, 2025
$60.60