MilliporeSigma
  • Characterization of cytochalasins from the endophytic Xylaria sp. and their biological functions.
Characterization of cytochalasins from the endophytic Xylaria sp. and their biological functions.
Journal of agricultural and food chemistry (2014-10-29)
Qiang Zhang, Jian Xiao, Qing-Qing Sun, Jian-Chun Qin, Gennaro Pescitelli, Jin-Ming Gao
ABSTRACT
Bioassay-guided fractionation of the fermentation extract of Xylaria sp. XC-16, an endophyte from Toona sinensis led to the isolation of two new cytochalasans cytochalasin Z27, 1, and cytochalasin Z28, 2, along with three known compounds seco-cytochalasin E, 3, and cytochalasin Z18, 4, and cytochalasin E, 5. The structures of 1 and 2 were elucidated by spectroscopic and electronic circular dichroism methods. Compound 5 was shown to be potently cytotoxic against brine shrimp (LC50 = 2.79 μM), comparable to that of the positive agent toosendanin (LC50 = 4.03 μM), and also exhibited potential phytotoxic effects on Lactuca sativa and Raphanus sativus L. seedlings, which are higher than that of the positive control glyphosate. Additionally, the fungicidal effect of 2 against the phytopathogen Gibberella saubinetti was better than that of hymexazol. This is the first report of the three types of cytochalasins present in genus Xylaria. A structure-phytotoxicity activity relationship is also discussed.
Sigma-Aldrich
3-Hydroxy-5-methylisoxazole, ≥90%