MilliporeSigma
  • Regulatory role of Lactobacillus acidophilus on inflammation and gastric dysmotility in intestinal mucositis induced by 5-fluorouracil in mice.
Regulatory role of Lactobacillus acidophilus on inflammation and gastric dysmotility in intestinal mucositis induced by 5-fluorouracil in mice.
Cancer chemotherapy and pharmacology (2015-01-13)
Priscilla F C Justino, Luis F M Melo, Andre F Nogueira, Cecila M Morais, Walber O Mendes, Alvaro X Franco, Emmanuel P Souza, Ronaldo A Ribeiro, Marcellus H L P Souza, Pedro Marcos Gomes Soares
ABSTRACT
Lactobacillus acidophilus is widely used for gastrointestinal disorders, but its role in inflammatory conditions like in chemotherapy-induced mucositis is unclear. Here, we report the effect of L. acidophilus on 5-fluorouracil-induced (5-FU) intestinal mucositis in mice. Mice weighing 25-30 g (n = 8) were separated into three groups, saline, 5-FU, and 5-FU + L. acidophilus (5-FU-La) (16 × 10(9) CFU/kg). In the 5-FU-La group, L. acidophilus was administered concomitantly with 5-FU on the first day and alone for two additional days. Three days after the last administration of L. acidophilus, the animals were euthanized and the jejunum and ileum were removed for histopathological assessment and for evaluation of levels of myeloperoxidase activity, sulfhydryl groups, nitrite, and cytokines (TNF-α, IL-1β, CXCL-1, and IL-10). In addition, we investigated gastric emptying using spectrophotometry after feeding a 1.5-ml test meal by gavage and euthanasia. Data were submitted to ANOVA and Bonferroni's test, with the level of significance at p < 0.05. Intestinal mucositis induced by 5-FU significantly (p < 0.05) reduced the villus height-crypt depth ratio and GSH concentration and increased myeloperoxidase activity and the nitrite concentrations compared with the control group. Furthermore, 5-FU significantly (p < 0.05) increased cytokine (TNF-α, IL-1β, and CXCL-1) concentrations and decreased IL-10 concentrations compared with the control group. 5-FU also significantly (p < 0.05) delayed gastric emptying and gastrointestinal transit compared with the control group. All of these changes were significantly (p < 0.05) reversed by treatment with L. acidophilus. Lactobacillus acidophilus improves the inflammatory and functional aspects of intestinal mucositis induced by 5-FU.
Sigma-Aldrich
o-Dianisidine dihydrochloride, Suitable for use in glucose determination
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
100 mL
In Stock
Details...
$51.90
6 x 20 mL
Estimated to ship on March 21, 2025
$60.60
Sigma-Aldrich
Hydrogen peroxide solution, SAJ first grade, ≥30.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
100 mL
In Stock
Details...
$51.90
6 x 20 mL
Estimated to ship on March 21, 2025
$60.60
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
100 mL
In Stock
Details...
$51.90
6 x 20 mL
Estimated to ship on March 21, 2025
$60.60
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
100 mL
In Stock
Details...
$51.90
6 x 20 mL
Estimated to ship on March 21, 2025
$60.60
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
100 mL
In Stock
Details...
$51.90
6 x 20 mL
Estimated to ship on March 21, 2025
$60.60
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
100 mL
In Stock
Details...
$51.90
6 x 20 mL
Estimated to ship on March 21, 2025
$60.60
Sigma-Aldrich
o-Dianisidine dihydrochloride, ≥95%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
100 mL
In Stock
Details...
$51.90
6 x 20 mL
Estimated to ship on March 21, 2025
$60.60
Sigma-Aldrich
o-Dianisidine dihydrochloride, tablet, 10 mg substrate per tablet
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
100 mL
In Stock
Details...
$51.90
6 x 20 mL
Estimated to ship on March 21, 2025
$60.60
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
100 mL
In Stock
Details...
$51.90
6 x 20 mL
Estimated to ship on March 21, 2025
$60.60
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
100 mL
In Stock
Details...
$51.90
6 x 20 mL
Estimated to ship on March 21, 2025
$60.60
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
100 mL
In Stock
Details...
$51.90
6 x 20 mL
Estimated to ship on March 21, 2025
$60.60
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
100 mL
In Stock
Details...
$51.90
6 x 20 mL
Estimated to ship on March 21, 2025
$60.60
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
100 mL
In Stock
Details...
$51.90
6 x 20 mL
Estimated to ship on March 21, 2025
$60.60
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
100 mL
In Stock
Details...
$51.90
6 x 20 mL
Estimated to ship on March 21, 2025
$60.60
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
100 mL
In Stock
Details...
$51.90
6 x 20 mL
Estimated to ship on March 21, 2025
$60.60
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
100 mL
In Stock
Details...
$51.90
6 x 20 mL
Estimated to ship on March 21, 2025
$60.60
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
100 mL
In Stock
Details...
$51.90
6 x 20 mL
Estimated to ship on March 21, 2025
$60.60
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
100 mL
In Stock
Details...
$51.90
6 x 20 mL
Estimated to ship on March 21, 2025
$60.60
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
100 mL
In Stock
Details...
$51.90
6 x 20 mL
Estimated to ship on March 21, 2025
$60.60
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
100 mL
In Stock
Details...
$51.90
6 x 20 mL
Estimated to ship on March 21, 2025
$60.60