Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sign Into View Organizational & Contract Pricing Indomethacin, European Pharmacopoeia (EP) Reference Standard
Sign Into View Organizational & Contract Pricing USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Potassium hydrogen phthalate, BioXtra, ≥99.95%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sign Into View Organizational & Contract Pricing Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing USP
Indomethacin, United States Pharmacopeia (USP) Reference Standard
Sign Into View Organizational & Contract Pricing Supelco
Indomethacin, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sign Into View Organizational & Contract Pricing Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium hydroxide solution, 0.01 M
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium hydroxide, SAJ first grade, ≥95.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium hydroxide solution, 0.2 M
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium hydroxide solution, 6 M
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium hydroxide, JIS special grade, ≥96.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sign Into View Organizational & Contract Pricing Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sign Into View Organizational & Contract Pricing Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium hydroxide solution, 7 M
Sign Into View Organizational & Contract Pricing