MilliporeSigma
Overexpression of Nrdp1/FLRF sensitizes cells to oxidative stress.
Biochemical and biophysical research communications (2011-06-23)
An Zhou, Danmin Pan, Xiaoming Yang, Jianhua Zhou
ABSTRACT
Nrdp1 is a RING finger containing ubiquitin E3 ligase that interacts with and modulates activity of multiple proteins, including ErbB3 and Parkin, a causative protein for early onset recessive juvenile parkinsonism (AR-JP). To investigate the functions of Nrdp1, we have generated stable Tet-On inducible HEK293 cells that overexpress Flag-tagged full length Nrdp1, N-terminal Nrdp1 and C-terminal Nrdp1. We demonstrate that overexpression of full-length Nrdp1, not Nrdp1 N-terminus or Nrdp1 C-terminus in cultured HEK293 cells, inhibits cell growth. In addition, we have treated cells with hydroxynonenal (HNE), 6-hydroxydopamine (6-OHDA), and hydrogen peroxide (H(2)O(2)) at different concentrations. We have found that Nrdp1 overexpression sensitizes HEK293 cells to oxidative stressors in a dosage-dependent manner. Our data provide insights into understanding the potential role of Nrdp1 in cell growth, apoptosis and oxidative stress, and in the pathogenesis of Parkinson's disease.