MilliporeSigma
  • Characterization of poly(L-lactic acid) microspheres loaded with holmium acetylacetonate.
Characterization of poly(L-lactic acid) microspheres loaded with holmium acetylacetonate.
Biomaterials (2001-09-29)
J F Nijsen, M J van Steenbergen, H Kooijman, H Talsma, L M Kroon-Batenburg, M van De Weert, P P van Rijk, A De Witte, A D Van Schip, W E Hennink
ABSTRACT
Holmium-loaded PLLA microspheres are useful systems in radioembolization therapy of liver metastases because of their low density, biodegradability and favourable radiation characteristics. Neutron activated Ho-loaded microspheres showed a surprisingly low release of the relatively small holmium complex. In this paper factors responsible for this behaviour are investigated, in particular by the use of differential scanning calorimetry, scanning electron microscopy, infrared spectroscopy and X-ray diffraction. The holmium complex is soluble in PLLA up to 8% in films and 17% in microspheres. Interactions between carbonyl groups of PLLA, and the Ho-ion in the HoAcAc complex, explain very satisfactorily the high stability of holmium-loaded microspheres.
Sigma-Aldrich
Zinc sulfate heptahydrate, ACS reagent, 99%
Sigma-Aldrich
Zinc sulfate heptahydrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5%