MilliporeSigma
  • A polymorphism in CALHM1 influences Ca2+ homeostasis, Abeta levels, and Alzheimer's disease risk.
A polymorphism in CALHM1 influences Ca2+ homeostasis, Abeta levels, and Alzheimer's disease risk.
Cell (2008-07-01)
Ute Dreses-Werringloer, Jean-Charles Lambert, Valérie Vingtdeux, Haitian Zhao, Horia Vais, Adam Siebert, Ankit Jain, Jeremy Koppel, Anne Rovelet-Lecrux, Didier Hannequin, Florence Pasquier, Daniela Galimberti, Elio Scarpini, David Mann, Corinne Lendon, Dominique Campion, Philippe Amouyel, Peter Davies, J Kevin Foskett, Fabien Campagne, Philippe Marambaud
ABSTRACT
Alzheimer's disease (AD) is a genetically heterogeneous disorder characterized by early hippocampal atrophy and cerebral amyloid-beta (Abeta) peptide deposition. Using TissueInfo to screen for genes preferentially expressed in the hippocampus and located in AD linkage regions, we identified a gene on 10q24.33 that we call CALHM1. We show that CALHM1 encodes a multipass transmembrane glycoprotein that controls cytosolic Ca(2+) concentrations and Abeta levels. CALHM1 homomultimerizes, shares strong sequence similarities with the selectivity filter of the NMDA receptor, and generates a large Ca(2+) conductance across the plasma membrane. Importantly, we determined that the CALHM1 P86L polymorphism (rs2986017) is significantly associated with AD in independent case-control studies of 3404 participants (allele-specific OR = 1.44, p = 2 x 10(-10)). We further found that the P86L polymorphism increases Abeta levels by interfering with CALHM1-mediated Ca(2+) permeability. We propose that CALHM1 encodes an essential component of a previously uncharacterized cerebral Ca(2+) channel that controls Abeta levels and susceptibility to late-onset AD.