MilliporeSigma
  • Reduction in serum aquaporin-4 antibody titers during development of a tumor-like brain lesion in a patient with neuromyelitis optica: a serum antibody-consuming effect?
Reduction in serum aquaporin-4 antibody titers during development of a tumor-like brain lesion in a patient with neuromyelitis optica: a serum antibody-consuming effect?
Journal of neuropathology and experimental neurology (2015-02-11)
Fahmy Aboulenein-Djamshidian, Romana Höftberger, Patrick Waters, Wolfgang Krampla, Hans Lassmann, Herbert Budka, Angela Vincent, Wolfgang Kristoferitsch
ABSTRACT
Neuromyelitis optica (NMO) is an inflammatory demyelinating disease of the CNS with severe involvement of the optic nerve and spinal cord. Highly specific serum IgG autoantibodies (NMO-IgG) that react with aquaporin-4 (AQP4), the most abundant CNS water channel protein, are found in patients with NMO. However, in vivo evidence combining the results of AQP4 antibody serum levels and brain pathology is lacking. We report a patient with NMO whose AQP4 antibody levels decreased simultaneously with clinical deterioration caused by the development of a tumor-like brain lesion. In the seminecrotic biopsied brain lesion, there was activated complement complex, whereas only very scattered immunoreactivity to AQP4 protein was detectable. The decrease in serum AQP4 antibody levels and the loss of AQP4 in the tumor-like lesion could represent a "serum antibody-consuming effect" during lesion formation.