MilliporeSigma
  • Comparison of micro- and mesoporous inorganic materials in the uptake and release of the drug model fluorescein and its analogues.
Comparison of micro- and mesoporous inorganic materials in the uptake and release of the drug model fluorescein and its analogues.
Chemistry (Weinheim an der Bergstrasse, Germany) (2003-12-16)
Karen A Fisher, Katherine D Huddersman, M Joan Taylor
ABSTRACT
The uptake of the three species of the drug model fluorescein (fluorescein sodium salt (FNa), fluorescein free acid (F), and fluorescein diacetate (FDA)) by zeolite NaX and the mesoporous zeotype MCM-41 was investigated as well as their release rates into solutions at pH 7 and pH 4.5. UV/Vis analysis was carried out at a wavelength of 490 nm. Uptakes of the sodium salt of 9 % for zeolite X and 14 % for MCM suggest little penetration of the pores. The use of ethanol as the loading solvent for F resulted in little uptake for both zeolitic materials due to the successful competition of the ethanol for binding sites. Use of acetone (weaker proton acceptor) as loading solvent significantly improved the uptake of F to 17 % and 12 % for zeolite X and MCM, respectively, whilst the uptake of FDA in acetone increased still further to 22 % and 17 % for zeolite X and MCM, respectively. Generally there was a large initial release of the fluorescein analogues from the surface of the zeolites with very little further increase over time. The prescence of an esterase enzyme in the release medium of FDA tripled the release from MCM to 15 % but left the release from zeolite X unaffected at 6 %. The results obtained show that uptake of fluorescein and its analogues is dependent on the loading solvent used, the amount released is influenced by not only the solvent but the pH and the presence of enzymes in the release medium.
Sigma-Aldrich
Fluorescein (free acid), Dye content 95 %