MilliporeSigma
  • Action of thiazolidine-2-carboxylic acid, a proline analog, on protein synthesizing systems.
Action of thiazolidine-2-carboxylic acid, a proline analog, on protein synthesizing systems.
Biochimica et biophysica acta (1979-09-27)
V Busiello, M di Girolamo, C Cini, C De Marco
PMID385056
ABSTRACT
Thiazolidine-2-carboxylic acid, or beta-thiaproline, is a proline analog in which the beta methylene group of proline is substituted by a sulfur atom. It has been deomonstrated that beta-thiaproline is activated and transferred to tRNAPro by Escherichia coli and rat liver aminoacyl-tRNA synthetases, and inhibits proline incorporation into polypeptides in protein synthesizing systems from E. coli, rat liver or rabbit reticulocytes. In mammalian systems beta-thiaproline inhibits also leucine incorporation; in rabbit reticulocyte lysate it inhibits ribosome run-off. Both these effects may be explained by the fact that beta-thiaproline once incorporated into the growing polypeptide chain impairs its further elongation, as shown by experiments made with puromycin. All tests were performed in comparison with thiazolidine-4-carboxylic acid, or gamma-thiaproline, another proline analog having the gamma methylene group substituted by a sulfur atom; it was shown that in all the reactions studied both compounds act as competitive inhibitors of proline. Some differences in the effects of the two analogs have been evidenced: in almost all the reactions and mainly in the whole protein synthesizing systems, beta-thiaproline shows an higher inhibitory activity.
Sigma-Aldrich
Thiazolidine-2-carboxylic acid, 97%