MilliporeSigma
  • Catalyzed Reformatsky reactions with ethyl bromofluoroacetate for the synthesis of alpha-fluoro-beta-hydroxy acids.
Catalyzed Reformatsky reactions with ethyl bromofluoroacetate for the synthesis of alpha-fluoro-beta-hydroxy acids.
The Journal of organic chemistry (2002-01-05)
R Ocampo, W R Dolbier, K A Abboud, F Zuluaga
ABSTRACT
The presence of catalytic amounts of CeCl(3) improves yields and simplifies procedure in the Reformatsky reactions of ethyl bromofluoroacetate with aldehydes and ketones to generate diastereomeric mixtures of alpha-fluoro-beta-hydroxy esters, some of which can be separated by crystallization or column flash chromatography. Diastereomerically pure alpha-fluoro-beta-hydroxy acids are obtained by mild alkaline hydrolysis of the resolved alpha-fluoro-beta-hydroxy esters. Detailed NMR data of new alpha-fluoro-beta-hydroxy esters and alpha-fluoro-beta-hydroxy acids are also presented.