MilliporeSigma
  • The ALG-2 binding site in Sec31A influences the retention kinetics of Sec31A at the endoplasmic reticulum exit sites as revealed by live-cell time-lapse imaging.
The ALG-2 binding site in Sec31A influences the retention kinetics of Sec31A at the endoplasmic reticulum exit sites as revealed by live-cell time-lapse imaging.
Bioscience, biotechnology, and biochemistry (2010-09-14)
Hideki Shibata, Tatsutoshi Inuzuka, Haruna Yoshida, Hirofumi Sugiura, Ikuo Wada, Masatoshi Maki
ABSTRACT
ALG-2, a member of the penta-EF-hand protein family, interacts Ca²+-dependently with a COPII component, Sec31A. In this study, we first established HeLa cells stably expressing green fluorescent protein-fused ALG-2 (GFP-ALG-2) and red fluorescent protein-fused Sec31A (Sec31A-RFP). After inducing Ca²+-mobilization, the cytoplasmic distribution of GFP-ALG-2 changed from a diffuse to a punctate pattern, which extensively overlapped with the Sec31A-RFP-positive structures, indicating that ALG-2 is recruited to the endoplasmic reticulum exit sites (ERES) in living cells. Next, overlay experiments with biotin-labeled ALG-2 were done to dissect the ALG-2 binding site (ABS). They revealed that a sequence comprising amino acid residues 839-851 in the Pro-rich region was necessary and sufficient for direct binding to ALG-2. Finally, fluorescence recovery after photobleaching analysis indicated that the ABS deletion reduced the high-affinity population of Sec31A to the ERES, suggesting that the ABS is one of the key determinants of the retention kinetics of Sec31A at ERES.