MilliporeSigma
S100A10/p11: family, friends and functions.
Pflugers Archiv : European journal of physiology (2007-07-20)
Ursula Rescher, Volker Gerke
ABSTRACT
S100A10, also known as p11 or annexin 2 light chain, is a member of the S100 family of small, dimeric EF hand-type Ca(2+)-binding proteins that generally modulate cellular target proteins in response to intracellular Ca(2+) signals. In contrast to all other S100 proteins, S100A10 is Ca(2+) insensitive because of amino acid replacements in its Ca(2+)-binding loops that lock the protein in a permanently active state. Within cells, the majority of S100A10 resides in a tight heterotetrameric complex with the peripheral membrane-binding protein annexin A2 that directs the complex to specific target membranes, in particular the plasma membrane and the membrane of early endosomes. Several other Ca(2+)-independent interaction partners of S100A10 have been described in the recent past. Many of these interactions, which have been shown to be of functional significance for the respective partner, involve plasma membrane-resident proteins. In most of these cases, S100A10, probably residing in a complex with annexin A2, appears to regulate the intracellular trafficking of the respective target protein and thus its functional expression at the cell surface. In this paper, we review the current information on S100A10 protein interactions placing a particular emphasis on data that contribute to an understanding of the mechanistic basis of the S100A10 action. Based on these data, we propose that S100A10 functions as a linker tethering certain transmembrane proteins to annexin A2 thereby assisting their traffic to the plasma membrane and/or their firm anchorage at certain membrane sites.