MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Human selenium binding protein-1 (hSP56) is a negative regulator of HIF-1α and suppresses the malignant characteristics of prostate cancer cells.
Human selenium binding protein-1 (hSP56) is a negative regulator of HIF-1α and suppresses the malignant characteristics of prostate cancer cells.
BMB reports (2014-05-31)
Jee-Yeong Jeong, Jin-Rong Zhou, Chong Gao, Laurie Feldman, Arthur J Sytkowski
ABSTRACT
In the present study, we demonstrate that ectopic expression of 56-kDa human selenium binding protein-1 (hSP56) in PC-3 cells that do not normally express hSP56 results in a marked inhibition of cell growth in vitro and in vivo. Down-regulation of hSP56 in LNCaP cells that normally express hSP56 results in enhanced anchorage-independent growth. PC-3 cells expressing hSP56 exhibit a significant reduction of hypoxia inducible protein (HIF)-1α protein levels under hypoxic conditions without altering HIF-1α mRNA (HIF1A) levels. Taken together, our findings strongly suggest that hSP56 plays a critical role in prostate cells by mechanisms including negative regulation of HIF-1α, thus identifying hSP56 as a candidate anti-oncogene product.