MilliporeSigma
  • NeuroD1/beta2 contributes to cell-specific transcription of the proopiomelanocortin gene.
NeuroD1/beta2 contributes to cell-specific transcription of the proopiomelanocortin gene.
Molecular and cellular biology (1997-10-29)
G Poulin, B Turgeon, J Drouin
ABSTRACT
NeuroD1/beta2 is a basic helix-loop-helix (bHLH) factor expressed in the endocrine cells of the pancreas and in a subset of neurons as they undergo terminal differentiation. We now show that NeuroD1 is expressed in corticotroph cells of the pituitary gland and that it is involved in cell-specific transcription of the proopiomelanocortin (POMC) gene. It was previously shown that corticotroph-specific POMC transcription depends in part on the action of cell-restricted bHLH factors that were characterized as the CUTE (corticotroph upstream transcription element) (M. Therrien and J. Drouin, Mol. Cell. Biol. 13:2342-2353, 1993) complexes. We now demonstrate that these complexes contain NeuroD1 in association with various ubiquitous bHLH dimerization partners. The NeuroD1-containing heterodimers specifically recognize and activate transcription from the POMC promoter E box that confers transcriptional specificity. Interestingly, the NeuroD1 heterodimers activate transcription in synergy with Ptx1, a Bicoid-related homeodomain protein, which also contributes to corticotroph specificity of POMC transcription. In the adult pituitary gland, NeuroD1 transcripts are detected in POMC-expressing corticotroph cells. Taken together with the restricted pattern of Ptx1 expression, these results suggest that these two factors establish the basis of a combinatorial code for the program of corticotroph-specific gene expression.