MilliporeSigma
  • ITI-H4, as a biomarker in the serum of recurrent pregnancy loss (RPL) patients.
ITI-H4, as a biomarker in the serum of recurrent pregnancy loss (RPL) patients.
Molecular bioSystems (2011-02-19)
Myung-Sun Kim, Bon-Hee Gu, Sangjin Song, Bum-Chae Choi, Dong-Hyun Cha, Kwang-Hyun Baek
ABSTRACT
Recurrent pregnancy loss (RPL) is defined as at least three pregnancy losses in series prior to the 20-28 weeks of pregnancy. There are several etiological factors associated with immunology, anatomy, endocrinology, genetic, infection, chromosomal abnormalities, and environmental factors contributing to the condition. The aim of this study was to identify RPL associated factors in human blood using proteomics. Since it is difficult to obtain tissues or follicular fluids, we used blood samples from normal and RPL patients to conduct a comparative proteomic study. Three RPL blood samples and one cocktailed blood sample from 3 normal women were used. We performed 2-DE and selected spots were analyzed with MALDI-TOF/MS. In the three RPL blood samples, 2-DE analysis revealed 549, 563 and 533 spots to be differentially expressed, respectively. Through a comparative analysis between the control and RPL, 21 spots were shown to be differentially expressed. Of these, 5 proteins were confirmed by Western blot analysis. One of these proteins, ITI-H4 (inter-α trypsin inhibitor-heavy chain 4), was weakly expressed at a molecular weight of 120 kDa, but was highly expressed at a modified molecular weight of 36 kDa in RPL patients. These findings suggest that ITI-H4 expression may be used as a biomarker, which could facilitate the development of novel diagnostic and therapeutic tools.