MilliporeSigma
  • Down-regulation of Z39Ig on macrophages by IFN-gamma in patients with chronic HBV infection.
Down-regulation of Z39Ig on macrophages by IFN-gamma in patients with chronic HBV infection.
Clinical immunology (Orlando, Fla.) (2010-04-20)
Sheng Guo, Chengying Yang, Feng Mei, Shengxi Wu, Na Luo, Lei Fei, Yongwen Chen, Yuzhang Wu
ABSTRACT
Co-inhibitory signals from the B7 superfamily have been demonstrated to induce T cell dysfunction in chronic HBV infection (CHB). However, the expression and function of Z39Ig, a new inhibitor of the B7 superfamily, is still unclear in CHB. Here immunohistochemical staining showed that Z39Ig was restricted to macrophages and that its level was decreased significantly in CHB patients compared to healthy controls. Moreover, reduced Z39Ig expression was positively correlated with plasma HBV load but was inversely related to serum alanine aminotransaminase levels. Further, Z39Ig mRNA had a negative relation to IFN-gamma in vivo, and IFN-gamma also down-regulated Z39Ig expression on monocyte-derived macrophages (MDMs) in a time- and dose-dependent manner in vitro. Interestingly, Z39Ig expression on MDMs was restored when IFN-gamma neutralizing antibodies were added to the T cell/MDM co-culture system, indicating that the IFN-gamma derived from activated-T cells may contribute to the reduction of Z39Ig in the CHB environment. Our results suggest that T cells can opposite T cell hyporesponsiveness through dampening Z39Ig inhibitory signals from macrophages and thus maintain their anti-viral function in CHB. Therefore, decreasing Z39Ig signals from macrophages could contribute to CHB clinical therapy.