MilliporeSigma
  • Myocyte enhancer factor 2A and 2D undergo phosphorylation and caspase-mediated degradation during apoptosis of rat cerebellar granule neurons.
Myocyte enhancer factor 2A and 2D undergo phosphorylation and caspase-mediated degradation during apoptosis of rat cerebellar granule neurons.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2001-08-23)
M Li, D A Linseman, M P Allen, M K Meintzer, X Wang, T Laessig, M E Wierman, K A Heidenreich
ABSTRACT
Myocyte enhancer factor 2 (MEF2) proteins are important regulators of gene expression during the development of skeletal, cardiac, and smooth muscle. MEF2 proteins are also present in brain and recently have been implicated in neuronal survival and differentiation. In this study we examined the cellular mechanisms regulating the activity of MEF2s during apoptosis of cultured cerebellar granule neurons, an established in vitro model for studying depolarization-dependent neuronal survival. All four MEF2 isoforms (A, B, C, and D) were detected by immunoblot analysis in cerebellar granule neurons. Endogenous MEF2A and MEF2D, but not MEF2B or MEF2C, were phosphorylated with the induction of apoptosis. The putative sites that were phosphorylated during apoptosis are functionally distinct from those previously reported to enhance MEF2 transcription. The increased phosphorylation of MEF2A and MEF2D was followed by decreased DNA binding, reduced transcriptional activity, and caspase-dependent cleavage to fragments containing N-terminal DNA binding domains and C-terminal transactivation domains. Expression of the highly homologous N terminus of MEF2A (1-131 amino acids) antagonized the transcriptional activity and prosurvival effects of a constitutively active mutant of MEF2D (MEF2D-VP16). We conclude that MEF2A and MEF2D are prosurvival factors with high transcriptional activity in postmitotic cerebellar granule neurons. When these neurons are induced to undergo apoptosis by lowering extracellular potassium, MEF2A and MEF2D are phosphorylated, followed by decreased DNA binding and cleavage by a caspase-sensitive pathway to N-terminal fragments lacking the transactivation domains. The degradation of MEF2D and MEF2A and the generation of MEF2 fragments that have the potential to act as dominant-inactive transcription factors lead to apoptotic cell death.