- 4-Cyano-3-fluoro-phenyl 4-(hexa-dec-yl-oxy)benzoate.
4-Cyano-3-fluoro-phenyl 4-(hexa-dec-yl-oxy)benzoate.
Acta crystallographica. Section E, Structure reports online (2014-04-26)
M K Usha, H T Srinivas, Rajni Kant, Vivek K Gupta, D Revannasiddaiah
PMID24764965
ABSTRACT
In the title compound, C30H40FNO3, the dihedral angle between the benzene rings is 57.76 (7)°. The alkyl chain adopts an all-trans conformation. In the crystal, mol-ecules are linked by pairs of C-H⋯O hydrogen bonds, forming inversion dimers.