MilliporeSigma
  • Interactions of partial LSD analogs with behavioral disrupting effects of LSD and DMT in the rat.
Interactions of partial LSD analogs with behavioral disrupting effects of LSD and DMT in the rat.
European journal of pharmacology (1978-01-01)
B Kovacic, L J Wang Lu, D Ruffing, E F Domino
PMID271075
ABSTRACT
Adult male Holtzman rats were trained to barpress on a schedule whereby every fourth press earned a reward of 0.01 ml of sugar-sweetened milk (FR4). After an i.p. injection of LSD (0.1 mg/kg) or DMT (3.2 or 10 mg/kg) such barpressing is abolished completely and resumed, usually within an hour, at a rate near the preinjection control rate of pressing. It continues at a steady, uninterrupted pace until the animals are removed from the operant chamber one-half hour later. A series of N,N-diethylnipecotamide derivatives were synthesized and tested for their ability to modify the disruptive effect of these hallucinogens. N,N-diethylbutyramide (DBA) and 1-methyl-1,2,5,6-tetrahydropyridine-3-(N,N-diethylcarboxamide) (THPC) were also tested. Pretreatment with a single i.p. injection of any of these compounds (5--40 mg/kg) either had no effect on or else prolonged the duration of hallucinogen-induced cessation of barpressing.