MilliporeSigma
  • Combined kinetic studies and computational analysis on kojic acid analogous as tyrosinase inhibitors.
Combined kinetic studies and computational analysis on kojic acid analogous as tyrosinase inhibitors.
Molecules (Basel, Switzerland) (2014-07-09)
Carlyle Ribeiro Lima, José Rogério A Silva, Erica de Tássia Carvalho Cardoso, Edilene O Silva, Jerônimo Lameira, José Luiz Martins do Nascimento, Davi do Socorro Barros Brasil, Cláudio N Alves
ABSTRACT
Tyrosinase is a key enzyme in melanin synthesis and widely distributed in plants and animals tissues. In mammals, this enzyme is related to pigment production, involved in wound healing, primary immune response and it can also contribute to catecholamines synthesis in the brain. Consequently, tyrosinase enzyme represents an attractive and selective target in the field of the medicine, cosmetics and bio-insecticides. In this paper, experimental kinetics and computational analysis were used to study the inhibition of tyrosinase by analogous of Kojic acid. The main interactions occurring between inhibitors-tyrosinase complexes and the influence of divalent cation (Cu2+) in enzymatic inhibition were investigated by using molecular docking, molecular dynamic simulations and electrostatic binding free energy by using the Linear Interaction Energy (LIE) method. The results showed that the electrostatic binding free energy are correlated with values of constant inhibition (r2 = 0.97).Thus, the model obtained here could contribute to future studies of this important system and, therefore, eventually facilitate development of tyrosinase inhibitors.
Sigma-Aldrich
2-Ethyl-3-hydroxy-4H-pyran-4-one, 99%
Pricing and availability is not currently available.