MilliporeSigma
  • Homogeneous supported synthesis using ionic liquid supports: tunable separation properties.
Homogeneous supported synthesis using ionic liquid supports: tunable separation properties.
The Journal of organic chemistry (2005-03-25)
Scott T Handy, Maurice Okello
ABSTRACT
[structure: see text] We report a homogeneous supported version of Koser's salt based on a room-temperature ionic liquid (RTIL) support. By altering the nature of the RTIL, a material was developed that was stable, recyclable, and readily separable from the tosyloxylated ketone products just by using variations in solvent polarity. A similar approach should be applicable to a wide range of supported catalysts and reagents.