MilliporeSigma
  • Intracavernous delivery of clonal mesenchymal stem cells restores erectile function in a mouse model of cavernous nerve injury.
Intracavernous delivery of clonal mesenchymal stem cells restores erectile function in a mouse model of cavernous nerve injury.
The journal of sexual medicine (2013-11-21)
Ji-Kan Ryu, Da-Ham Kim, Kang Moon Song, Tacghee Yi, Jun-Kyu Suh, Sun U Song
ABSTRACT
Recently, much attention has focused on stem cell therapy; bone marrow-derived stem cells (BMSCs) are one of the most studied mesenchymal stem cells used in the field of erectile dysfunction (ED). However, a major limitation for the clinical application of stem cell therapy is the heterogeneous nature of the isolated cells, which may cause different treatment outcomes. We investigated the effectiveness of mouse clonal BMSCs obtained from a single colony by using subfractionation culturing method (SCM) for erectile function in a mouse model of cavernous nerve injury (CNI). Twelve-week-old C57BL/6J mice were divided into four groups: sham operation group, bilateral CNI group receiving a single intracavernous (IC) injection of phosphate-buffered saline (20 μL) or clonal BMSCs (3 × 10(5) cells/20 μL), and receiving a single intraperitoneal (IP) injection of clonal BMSCs (3 × 10(5) cells/20 μL). The clonal BMSC line was analyzed for cell-surface epitopes by using fluorescence-activated cell sorting and for differentiation potential. Two weeks after CNI and treatment, erectile function was measured by electrically stimulating the cavernous nerve. The penis was harvested for histologic examinations and Western blot analysis. Clonal BMSCs expressed cell surface markers for mesenchymal stem cells and were capable of differentiating into several lineages, including adipogenic, osteogenic, and chondrogenic cells. Both IC and IP injections of clonal BMSCs significantly restored cavernous endothelial and smooth muscle content, and penile nNOS and neurofilament content in CNI mice. IC injection of clonal BMSCs induced significant recovery of erectile function, which reached 90-100% of the sham control values, whereas IP injection of clonal BMSCs partially restored erectile function. We established a homogeneous population of mouse clonal BMSCs using SCM; clonal BMSCs successfully restored erectile function in CNI mice. The homogeneous nature of clonal mesenchymal stem cells may allow their clinical applications.
Product Description
Dexamethasone, British Pharmacopoeia (BP) Assay Standard
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Selenous acid, 99.999% trace metals basis
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Selenous acid, 98%
Sigma-Aldrich
DL-Proline, ReagentPlus®, 99%
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, SAJ first grade, ≥35.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde solution, JIS special grade, 36.0-38.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde solution, 10%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade