MilliporeSigma
  • Effect of air classification and fermentation by Lactobacillus plantarum VTT E-133328 on faba bean (Vicia faba L.) flour nutritional properties.
Effect of air classification and fermentation by Lactobacillus plantarum VTT E-133328 on faba bean (Vicia faba L.) flour nutritional properties.
International journal of food microbiology (2014-12-03)
Rossana Coda, Leena Melama, Carlo Giuseppe Rizzello, José Antonio Curiel, Juhani Sibakov, Ulla Holopainen, Marjo Pulkkinen, Nesli Sozer
ABSTRACT
The effects of air classification and lactic acid bacteria fermentation on the reduction of anti-nutritional factors (vicine and convicine, trypsin inhibitor activity, condensed tannins and phytic acid) and in vitro protein and starch digestibility of faba bean flour were studied. Free amino acid (FAA) profile analysis was also carried out. Air classification allowed the separation of the flour into protein and starch rich fractions, showing different chemical compositions and microstructures. Lactobacillus plantarum growth and acidification in faba bean flour and its fractions were assessed. The anti-nutritional compounds were separated mostly to the fine protein-rich fraction. Fermentation caused the decrease of vicine and convicine contents by more than 91% and significantly reduced trypsin inhibitor activity and condensed tannins (by more than 40% in the protein-rich fraction). No significant (P>0.05) variation was observed for total phenols and phytic acid content. Fermentation increased the amount of FAA, especially of the essential amino acids and γ-aminobutyric acid, enhanced the in vitro protein digestibility and significantly lowered the hydrolysis index. This work showed that the combination of air classification and fermentation improved nutritional functionality of faba bean flour which could be utilized in various food applications.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Lactic acid, 85%, FCC
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Starch from potato, Powder
Sigma-Aldrich
Sodium hydroxide, SAJ first grade, ≥95.0%
Sigma-Aldrich
Sodium hydroxide solution, 0.2 M
Sigma-Aldrich
Acetic acid, SAJ first grade, ≥99.0%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Sodium hydroxide solution, 1 M
Sigma-Aldrich
Acetic acid, JIS special grade, ≥99.7%
Sigma-Aldrich
Acetic acid, 99.5-100.0%
Sigma-Aldrich
Sodium hydroxide solution, 7 M
Sigma-Aldrich
Sodium hydroxide, JIS special grade, ≥96.0%
Sigma-Aldrich
Sodium hydroxide solution, 6 M
Sigma-Aldrich
Acetic acid solution, 1 M, 1 N
Sigma-Aldrich
Sodium hydroxide solution, 0.01 M
Sigma-Aldrich
Acetic acid, ≥99.7%, suitable for amino acid analysis
Sigma-Aldrich
Sodium hydroxide solution, 0.1 M
Sigma-Aldrich
Sodium hydroxide solution, 4 M
Sigma-Aldrich
Sodium hydroxide solution, 0.05 M
Supelco
Lactic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Cycloheximide, ≥90% (HPLC)
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Supelco
γ-Aminobutyric acid, analytical standard
Sigma-Aldrich
Lactic acid, meets USP testing specifications
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%