MilliporeSigma
  • 1-[2,6-Dichloro-4-(trifluoro-meth-yl)phen-yl]-3,4-dimethyl-pyrano[2,3-c]pyrazol-6(1H)-one.
1-[2,6-Dichloro-4-(trifluoro-meth-yl)phen-yl]-3,4-dimethyl-pyrano[2,3-c]pyrazol-6(1H)-one.
Acta crystallographica. Section E, Structure reports online (2012-08-21)
Hoong-Kun Fun, Wan-Sin Loh, B K Sarojini, B J Mohan, B Narayana
ABSTRACT
In the title compound, C(15)H(9)Cl(2)F(3)N(2)O(2), the 1,6-dihydro-pyrano[2,3-c]pyrazole ring system is almost planar, with a maximum deviation of 0.0226 (14) Å, and forms a dihedral angle of 69.90 (6)° with the benzene ring. In the crystal, mol-ecules are linked into a helical chain along the c axis by C-H⋯O hydrogen bonds.