MilliporeSigma
  • Rapid and simple comparison of messenger RNA levels using real-time PCR.
Rapid and simple comparison of messenger RNA levels using real-time PCR.
Biological procedures online (2006-02-01)
Andrée-Anne Dussault, Marc Pouliot
ABSTRACT
Real-time polymerase chain reaction (PCR) constitutes a significant improvement over traditional end-point PCR, as it allows the quantification of starting amounts of nucleic acid templates, in real-time. However, quantification requires validation through numerous internal controls and standard curves. We describe in this paper a simple protocol which uses real-time PCR to compare mRNA levels of a gene of interest between different experimental conditions. Comparative real-time PCR can be a relatively low-cost method and does not require sequence-specific fluorescent reporters. Moreover, several genes from a set of experiments can be assessed in a single run. Thus, in addition to providing a comparative profile for the expression of a gene of interest, this method can also provide information regarding the relative abundance of different mRNA species.
Sigma-Aldrich
10X PCR Buffer without MgCl2, Optimized for routine PCR without MgCl2
Pricing and availability is not currently available.