MilliporeSigma
Apaf-1 and caspase-9 in p53-dependent apoptosis and tumor inhibition.
Science (New York, N.Y.) (1999-04-02)
M S Soengas, R M Alarcón, H Yoshida, A J Giaccia, R Hakem, T W Mak, S W Lowe
ABSTRACT
The ability of p53 to promote apoptosis in response to mitogenic oncogenes appears to be critical for its tumor suppressor function. Caspase-9 and its cofactor Apaf-1 were found to be essential downstream components of p53 in Myc-induced apoptosis. Like p53 null cells, mouse embryo fibroblast cells deficient in Apaf-1 and caspase-9, and expressing c-Myc, were resistant to apoptotic stimuli that mimic conditions in developing tumors. Inactivation of Apaf-1 or caspase-9 substituted for p53 loss in promoting the oncogenic transformation of Myc-expressing cells. These results imply a role for Apaf-1 and caspase-9 in controlling tumor development.