MilliporeSigma
  • A novel glycine decarboxylase gene mutation in an Indian family with nonketotic hyperglycinemia.
A novel glycine decarboxylase gene mutation in an Indian family with nonketotic hyperglycinemia.
Journal of child neurology (2013-01-26)
Jennifer M Love, Debra Prosser, Donald R Love, Krishna Prakash Chintakindi, Ashwin B Dalal, Shagun Aggarwal
ABSTRACT
Nonketotic hyperglycinemia is an inborn error of glycine metabolism. It manifests mostly as an acute encephalopathy in the neonatal period, although later, atypical presentations have also been reported. Mutations in 3 different genes have been implicated in nonketotic hyperglycinemia. Here we report a novel mutation, c.2296G>T (p.Gly766Cys), in exon 19 of the glycine decarboxylase (GLDC) gene (Refseq accession number NM_000170.2) in a consanguineous Indian couple with a history of 4 neonatal deaths.